Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101583373-101583644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLL
Entrez Gene ID
27343
Gene Name
polymerase (DNA) lambda
Gene Aliases
BETAN, POLKAPPA
UniGene
Hs.523230
Gene Chromosome Location
Chr.10: 101578882-101588270 on Build GRCh38
Transcripts
NM_001174084.1 NM_001174085.1 NM_001308382.1 NM_013274.3 NR_033406.1 XM_006717775.1 XM_006717776.1 XM_006717777.1 XM_011539650.1 XM_011539651.1 XM_011539652.1 XM_011539653.1 XM_011539654.1 XM_011539655.1 XM_011539656.1 XM_011539657.1 XM_011539659.1 XM_011539660.1 XM_011539662.1 XM_011539663.1 XM_011539664.1 XM_011539665.2 XM_011539666.1 XM_011539667.1 XM_017016084.1 XM_017016085.1 XM_017016086.1 XM_017016087.1 XM_017016088.1 XM_017016089.1 XM_017016090.1 XM_017016091.1 XM_017016092.1 XM_017016093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6419200 c.971A>T p.K324M Chr.10: 101583602-101583602 on Build GRCh38
5442255 c.733C>T p.L245F Chr.10: 101583564-101583564 on Build GRCh38
6365481 c.954G>T p.E318D Chr.10: 101583619-101583619 on Build GRCh38
6488076 c.985G>C p.E329Q Chr.10: 101583621-101583621 on Build GRCh38
4438592 c.655G>A p.E219K Chr.10: 101583642-101583642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995758361 Chr.10: 101583571-101583571 on Build GRCh38
rs761039339 Chr.10: 101583491-101583491 on Build GRCh38
rs749920056 Chr.10: 101583551-101583551 on Build GRCh38
rs368773624 Chr.10: 101583583-101583583 on Build GRCh38
rs779648703 Chr.10: 101583555-101583555 on Build GRCh38
rs371010766 Chr.10: 101583600-101583600 on Build GRCh38
rs565324718 Chr.10: 101583615-101583615 on Build GRCh38
rs908023111 Chr.10: 101583500-101583500 on Build GRCh38
rs766690469 Chr.10: 101583504-101583504 on Build GRCh38
rs751696073 Chr.10: 101583583-101583586 on Build GRCh38
rs201019235 Chr.10: 101583616-101583616 on Build GRCh38
rs776938693 Chr.10: 101583610-101583610 on Build GRCh38
rs1020858222 Chr.10: 101583517-101583517 on Build GRCh38
rs372176589 Chr.10: 101583419-101583419 on Build GRCh38
rs199820847 Chr.10: 101583485-101583485 on Build GRCh38
rs367651179 Chr.10: 101583613-101583613 on Build GRCh38
rs146197224 Chr.10: 101583557-101583557 on Build GRCh38
rs927499734 Chr.10: 101583452-101583452 on Build GRCh38
rs999272431 Chr.10: 101583554-101583554 on Build GRCh38
rs376406720 Chr.10: 101583461-101583461 on Build GRCh38
rs373803211 Chr.10: 101583605-101583605 on Build GRCh38
rs183248417 Chr.10: 101583511-101583511 on Build GRCh38
rs371199480 Chr.10: 101583466-101583466 on Build GRCh38
rs201742864 Chr.10: 101583609-101583609 on Build GRCh38
rs760809916 Chr.10: 101583606-101583606 on Build GRCh38
rs148400497 Chr.10: 101583580-101583580 on Build GRCh38
rs542569736 Chr.10: 101583520-101583520 on Build GRCh38
rs768509874 Chr.10: 101583568-101583568 on Build GRCh38
rs370688403 Chr.10: 101583540-101583541 on Build GRCh38
rs778506125 Chr.10: 101583579-101583579 on Build GRCh38
rs980359253 Chr.10: 101583449-101583449 on Build GRCh38
rs754298053 Chr.10: 101583505-101583505 on Build GRCh38
rs780744781 Chr.10: 101583550-101583550 on Build GRCh38
rs554844269 Chr.10: 101583552-101583552 on Build GRCh38
rs143224710 Chr.10: 101583508-101583508 on Build GRCh38
rs758762395 Chr.10: 101583527-101583527 on Build GRCh38
rs560893212 Chr.10: 101583574-101583574 on Build GRCh38

View Full Product Details