Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68900465-68900690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL21
Entrez Gene ID
219927
Gene Name
mitochondrial ribosomal protein L21
Gene Aliases
L21mt, MRP-L21
UniGene
Hs.503047
Gene Chromosome Location
Chr.11: 68891276-68903835 on Build GRCh38
Transcripts
NM_181514.1 NM_181515.1 XM_005273823.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6321762 c.124C>A p.Q42K Chr.11: 68900570-68900570 on Build GRCh38
5373411 c.92C>T p.S31F Chr.11: 68900602-68900602 on Build GRCh38
3375933 c.130A>G p.T44A Chr.11: 68900564-68900564 on Build GRCh38
3452536 c.115T>C p.F39L Chr.11: 68900579-68900579 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759738566 Chr.11: 68900541-68900541 on Build GRCh38
rs763482916 Chr.11: 68900548-68900548 on Build GRCh38
rs541342052 Chr.11: 68900590-68900590 on Build GRCh38
rs765533988 Chr.11: 68900544-68900544 on Build GRCh38
rs768993754 Chr.11: 68900639-68900639 on Build GRCh38
rs754802573 Chr.11: 68900582-68900582 on Build GRCh38
rs370356485 Chr.11: 68900645-68900645 on Build GRCh38
rs766946282 Chr.11: 68900652-68900652 on Build GRCh38
rs771141783 Chr.11: 68900589-68900589 on Build GRCh38
rs576394802 Chr.11: 68900512-68900512 on Build GRCh38
rs755730762 Chr.11: 68900576-68900576 on Build GRCh38
rs753008820 Chr.11: 68900514-68900526 on Build GRCh38
rs776787818 Chr.11: 68900537-68900537 on Build GRCh38
rs146932385 Chr.11: 68900584-68900584 on Build GRCh38
rs776793956 Chr.11: 68900658-68900658 on Build GRCh38
rs574307329 Chr.11: 68900524-68900524 on Build GRCh38
rs137864766 Chr.11: 68900585-68900585 on Build GRCh38
rs756569873 Chr.11: 68900629-68900631 on Build GRCh38
rs774903036 Chr.11: 68900644-68900644 on Build GRCh38
rs768661882 Chr.11: 68900519-68900519 on Build GRCh38
rs753712080 Chr.11: 68900579-68900579 on Build GRCh38
rs756815802 Chr.11: 68900560-68900560 on Build GRCh38
rs767047685 Chr.11: 68900569-68900569 on Build GRCh38
rs376576401 Chr.11: 68900577-68900577 on Build GRCh38
rs749192845 Chr.11: 68900501-68900501 on Build GRCh38
rs775875071 Chr.11: 68900545-68900545 on Build GRCh38
rs373501945 Chr.11: 68900529-68900529 on Build GRCh38
rs922203046 Chr.11: 68900655-68900655 on Build GRCh38
rs770017430 Chr.11: 68900606-68900606 on Build GRCh38
rs764625555 Chr.11: 68900555-68900555 on Build GRCh38
rs763356692 Chr.11: 68900636-68900636 on Build GRCh38
rs750045430 Chr.11: 68900575-68900575 on Build GRCh38
rs774403716 Chr.11: 68900520-68900520 on Build GRCh38
rs752038568 Chr.11: 68900557-68900557 on Build GRCh38

View Full Product Details