Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 134965160-134965433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBS1L
Entrez Gene ID
10767
Gene Name
HBS1 like translational GTPase
Gene Aliases
EF-1a, ERFS, HBS1, HSPC276, eRF3c
UniGene
Hs.378532
Gene Chromosome Location
Chr.6: 134960378-135054898 on Build GRCh38
Transcripts
NM_001145158.1 NM_006620.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1073591 c.2053T>G p.*685G Chr.6: 134965281-134965281 on Build GRCh38
4828018 c.2044-1G>A p.? Chr.6: 134965291-134965291 on Build GRCh38
6677823 c.2044-1G>T p.? Chr.6: 134965291-134965291 on Build GRCh38
1440616 c.2045T>C p.I682T Chr.6: 134965289-134965289 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs79041205 Chr.6: 134965311-134965312 on Build GRCh38
rs753092593 Chr.6: 134965300-134965301 on Build GRCh38
rs778867731 Chr.6: 134965267-134965267 on Build GRCh38
rs766251088 Chr.6: 134965232-134965232 on Build GRCh38
rs747148717 Chr.6: 134965303-134965303 on Build GRCh38
rs558533235 Chr.6: 134965301-134965302 on Build GRCh38
rs759137380 Chr.6: 134965337-134965337 on Build GRCh38
rs770886967 Chr.6: 134965323-134965323 on Build GRCh38
rs757762224 Chr.6: 134965288-134965288 on Build GRCh38
rs949235161 Chr.6: 134965203-134965203 on Build GRCh38
rs753513636 Chr.6: 134965271-134965271 on Build GRCh38
rs758995716 Chr.6: 134965301-134965301 on Build GRCh38
rs13199205 Chr.6: 134965367-134965367 on Build GRCh38
rs767241791 Chr.6: 134965270-134965270 on Build GRCh38
rs762691252 Chr.6: 134965318-134965318 on Build GRCh38
rs879067139 Chr.6: 134965387-134965387 on Build GRCh38
rs758819323 Chr.6: 134965301-134965303 on Build GRCh38
rs202046751 Chr.6: 134965300-134965300 on Build GRCh38
rs773008583 Chr.6: 134965317-134965317 on Build GRCh38
rs776687374 Chr.6: 134965300-134965300 on Build GRCh38
rs901495733 Chr.6: 134965351-134965351 on Build GRCh38
rs748316339 Chr.6: 134965308-134965308 on Build GRCh38
rs371072942 Chr.6: 134965304-134965304 on Build GRCh38
rs746066187 Chr.6: 134965339-134965339 on Build GRCh38
rs372618200 Chr.6: 134965259-134965259 on Build GRCh38
rs780381803 Chr.6: 134965302-134965302 on Build GRCh38
rs953599265 Chr.6: 134965384-134965384 on Build GRCh38
rs769940089 Chr.6: 134965312-134965312 on Build GRCh38
rs779423709 Chr.6: 134965298-134965298 on Build GRCh38
rs796306431 Chr.6: 134965312-134965312 on Build GRCh38
rs1028634175 Chr.6: 134965379-134965379 on Build GRCh38
rs879191684 Chr.6: 134965302-134965302 on Build GRCh38
rs35078158 Chr.6: 134965301-134965301 on Build GRCh38
rs569539618 Chr.6: 134965368-134965368 on Build GRCh38
rs776967433 Chr.6: 134965305-134965305 on Build GRCh38
rs1052649456 Chr.6: 134965343-134965343 on Build GRCh38
rs546589091 Chr.6: 134965213-134965213 on Build GRCh38
rs780475312 Chr.6: 134965389-134965389 on Build GRCh38
rs556264662 Chr.6: 134965302-134965303 on Build GRCh38
rs377434508 Chr.6: 134965276-134965276 on Build GRCh38
rs766503035 Chr.6: 134965282-134965286 on Build GRCh38

View Full Product Details