Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50510129-50510358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JOSD2
Entrez Gene ID
126119
Gene Name
Josephin domain containing 2
Gene Aliases
SBBI54
UniGene
Hs.467151
Gene Chromosome Location
Chr.19: 50505997-50511355 on Build GRCh38
Transcripts
NM_001270639.1 NM_001270640.1 NM_001270641.1 NM_001270686.1 NM_138334.3 XM_011526434.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
713293 c.129C>T p.A43A Chr.19: 50510303-50510303 on Build GRCh38
999748 c.146G>A p.R49K Chr.19: 50510286-50510286 on Build GRCh38
4897689 c.107A>C p.Q36P Chr.19: 50510325-50510325 on Build GRCh38
6689681 c.93C>T p.N31N Chr.19: 50510339-50510339 on Build GRCh38
4523556 c.121G>A p.E41K Chr.19: 50510311-50510311 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199948323 Chr.19: 50510332-50510332 on Build GRCh38
rs945046787 Chr.19: 50510177-50510177 on Build GRCh38
rs553214820 Chr.19: 50510178-50510178 on Build GRCh38
rs556493192 Chr.19: 50510265-50510265 on Build GRCh38
rs761401490 Chr.19: 50510276-50510276 on Build GRCh38
rs188064442 Chr.19: 50510267-50510267 on Build GRCh38
rs756222333 Chr.19: 50510297-50510297 on Build GRCh38
rs200968884 Chr.19: 50510333-50510333 on Build GRCh38
rs773968994 Chr.19: 50510273-50510273 on Build GRCh38
rs757604874 Chr.19: 50510318-50510318 on Build GRCh38
rs1049789545 Chr.19: 50510192-50510194 on Build GRCh38
rs889342971 Chr.19: 50510198-50510198 on Build GRCh38
rs554914569 Chr.19: 50510311-50510311 on Build GRCh38
rs943542642 Chr.19: 50510214-50510214 on Build GRCh38
rs1000389631 Chr.19: 50510211-50510211 on Build GRCh38
rs772087921 Chr.19: 50510242-50510242 on Build GRCh38
rs377312039 Chr.19: 50510302-50510302 on Build GRCh38
rs775288583 Chr.19: 50510251-50510251 on Build GRCh38
rs764069996 Chr.19: 50510294-50510294 on Build GRCh38
rs893115715 Chr.19: 50510277-50510277 on Build GRCh38
rs571175687 Chr.19: 50510213-50510213 on Build GRCh38
rs764844630 Chr.19: 50510279-50510279 on Build GRCh38
rs753496687 Chr.19: 50510270-50510270 on Build GRCh38
rs1029751783 Chr.19: 50510308-50510308 on Build GRCh38
rs142878293 Chr.19: 50510257-50510258 on Build GRCh38
rs370080842 Chr.19: 50510286-50510286 on Build GRCh38
rs781751120 Chr.19: 50510291-50510291 on Build GRCh38
rs779247091 Chr.19: 50510326-50510326 on Build GRCh38
rs903478660 Chr.19: 50510188-50510188 on Build GRCh38
rs1036734260 Chr.19: 50510176-50510176 on Build GRCh38
rs755291936 Chr.19: 50510289-50510289 on Build GRCh38
rs142440887 Chr.19: 50510184-50510184 on Build GRCh38
rs538269597 Chr.19: 50510229-50510229 on Build GRCh38
rs1055056376 Chr.19: 50510225-50510225 on Build GRCh38
rs1040483033 Chr.19: 50510218-50510218 on Build GRCh38

View Full Product Details