Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 42097307-42097568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GXYLT1
Entrez Gene ID
283464
Gene Name
glucoside xylosyltransferase 1
Gene Aliases
GLT8D3
UniGene
Hs.259347
Gene Chromosome Location
Chr.12: 42081845-42144871 on Build GRCh38
Transcripts
NM_001099650.1 NM_173601.1 XM_017019211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929396 c.1103G>A p.G368D Chr.12: 42097500-42097500 on Build GRCh38
2066521 c.1018C>T p.H340Y Chr.12: 42097492-42097492 on Build GRCh38
3811989 c.1024G>A p.D342N Chr.12: 42097486-42097486 on Build GRCh38
5929398 c.1102G>A p.G368S Chr.12: 42097501-42097501 on Build GRCh38
3811988 c.1117G>A p.D373N Chr.12: 42097486-42097486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758847983 Chr.12: 42097518-42097518 on Build GRCh38
rs751768814 Chr.12: 42097532-42097532 on Build GRCh38
rs758875099 Chr.12: 42097416-42097417 on Build GRCh38
rs79724807 Chr.12: 42097530-42097530 on Build GRCh38
rs141417516 Chr.12: 42097526-42097528 on Build GRCh38
rs747038768 Chr.12: 42097414-42097414 on Build GRCh38
rs377269156 Chr.12: 42097415-42097415 on Build GRCh38
rs374180059 Chr.12: 42097409-42097409 on Build GRCh38
rs571276787 Chr.12: 42097393-42097393 on Build GRCh38
rs1051424290 Chr.12: 42097504-42097504 on Build GRCh38
rs772963353 Chr.12: 42097477-42097477 on Build GRCh38
rs374749531 Chr.12: 42097540-42097540 on Build GRCh38
rs957445129 Chr.12: 42097359-42097359 on Build GRCh38
rs980971519 Chr.12: 42097347-42097347 on Build GRCh38
rs764193650 Chr.12: 42097529-42097529 on Build GRCh38
rs751614002 Chr.12: 42097417-42097417 on Build GRCh38
rs768731656 Chr.12: 42097455-42097455 on Build GRCh38
rs755446063 Chr.12: 42097433-42097433 on Build GRCh38
rs778115867 Chr.12: 42097410-42097410 on Build GRCh38
rs982080558 Chr.12: 42097392-42097392 on Build GRCh38
rs180977321 Chr.12: 42097366-42097366 on Build GRCh38
rs759977286 Chr.12: 42097497-42097497 on Build GRCh38
rs143025715 Chr.12: 42097453-42097453 on Build GRCh38
rs548843764 Chr.12: 42097363-42097363 on Build GRCh38
rs774219341 Chr.12: 42097459-42097459 on Build GRCh38
rs73128009 Chr.12: 42097502-42097502 on Build GRCh38
rs73280063 Chr.12: 42097388-42097388 on Build GRCh38
rs771608825 Chr.12: 42097476-42097476 on Build GRCh38
rs910507725 Chr.12: 42097435-42097435 on Build GRCh38
rs201958690 Chr.12: 42097485-42097485 on Build GRCh38
rs748038957 Chr.12: 42097466-42097466 on Build GRCh38
rs199516137 Chr.12: 42097499-42097499 on Build GRCh38
rs781456325 Chr.12: 42097450-42097450 on Build GRCh38
rs753117564 Chr.12: 42097407-42097407 on Build GRCh38
rs200393853 Chr.12: 42097487-42097487 on Build GRCh38
rs781562409 Chr.12: 42097531-42097533 on Build GRCh38
rs941108560 Chr.12: 42097461-42097461 on Build GRCh38
rs776140282 Chr.12: 42097495-42097495 on Build GRCh38
rs1049224865 Chr.12: 42097420-42097420 on Build GRCh38
rs200853648 Chr.12: 42097533-42097533 on Build GRCh38
rs765534902 Chr.12: 42097394-42097394 on Build GRCh38
rs186161087 Chr.12: 42097390-42097390 on Build GRCh38

View Full Product Details