Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 71878368-71878617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100129216
Entrez Gene ID
100129216
Gene Name
beta-defensin 131-like
Gene Aliases
-
UniGene
Hs.740246
Gene Chromosome Location
Chr.11: 71878453-71884561 on Build GRCh38
Transcripts
NM_001242853.1 NR_029192.1

Gene Symbol
LOC100133315
Entrez Gene ID
100133315
Gene Name
transient receptor potential cation channel, subfamily C, member 2-like
Gene Aliases
-
UniGene
Hs.150324
Gene Chromosome Location
Chr.11: 71865509-71928447 on Build GRCh38
Transcripts
NM_001242853.1 NR_029192.1

SNP Detail

SNP ID Chromosome Location
rs750719846 Chr.11: 71878491-71878491 on Build GRCh38
rs972930615 Chr.11: 71878517-71878517 on Build GRCh38
rs759409819 Chr.11: 71878444-71878444 on Build GRCh38
rs772776606 Chr.11: 71878415-71878415 on Build GRCh38
rs188359057 Chr.11: 71878547-71878547 on Build GRCh38
rs771770108 Chr.11: 71878493-71878493 on Build GRCh38
rs760518098 Chr.11: 71878433-71878433 on Build GRCh38
rs769041318 Chr.11: 71878471-71878471 on Build GRCh38
rs756508872 Chr.11: 71878459-71878459 on Build GRCh38
rs757597000 Chr.11: 71878453-71878453 on Build GRCh38
rs765098183 Chr.11: 71878549-71878549 on Build GRCh38
rs775479347 Chr.11: 71878554-71878554 on Build GRCh38
rs753763774 Chr.11: 71878452-71878452 on Build GRCh38
rs776655638 Chr.11: 71878532-71878532 on Build GRCh38
rs931161337 Chr.11: 71878561-71878561 on Build GRCh38
rs766354919 Chr.11: 71878437-71878437 on Build GRCh38
rs780639941 Chr.11: 71878462-71878462 on Build GRCh38
rs764366512 Chr.11: 71878445-71878445 on Build GRCh38
rs545212655 Chr.11: 71878508-71878508 on Build GRCh38
rs970296990 Chr.11: 71878454-71878454 on Build GRCh38
rs112890198 Chr.11: 71878395-71878395 on Build GRCh38
rs770736140 Chr.11: 71878522-71878522 on Build GRCh38
rs749564200 Chr.11: 71878464-71878464 on Build GRCh38
rs563480732 Chr.11: 71878546-71878546 on Build GRCh38
rs779288967 Chr.11: 71878487-71878487 on Build GRCh38
rs772652044 Chr.11: 71878500-71878500 on Build GRCh38
rs747793994 Chr.11: 71878488-71878488 on Build GRCh38
rs781403486 Chr.11: 71878457-71878457 on Build GRCh38
rs765541430 Chr.11: 71878465-71878465 on Build GRCh38

View Full Product Details