Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23753634-23753905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMA3
Entrez Gene ID
3909
Gene Name
laminin subunit alpha 3
Gene Aliases
BM600, E170, LAMNA, LOCS
UniGene
Hs.436367
Gene Chromosome Location
Chr.18: 23689443-23955066 on Build GRCh38
Transcripts
NM_001127717.2 NM_001302996.1 NM_198129.2 NR_130106.1 XM_011525978.2 XM_011525979.2 XM_011525980.2 XM_011525981.2 XM_011525982.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756097 c.891G>A p.G297G Chr.18: 23753756-23753756 on Build GRCh38
5818391 c.928A>G p.I310V Chr.18: 23753793-23753793 on Build GRCh38
1388046 c.947+1G>A p.? Chr.18: 23753813-23753813 on Build GRCh38
5387890 c.873G>A p.K291K Chr.18: 23753738-23753738 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781210954 Chr.18: 23753726-23753726 on Build GRCh38
rs773855908 Chr.18: 23753768-23753768 on Build GRCh38
rs545483100 Chr.18: 23753705-23753705 on Build GRCh38
rs530283690 Chr.18: 23753854-23753854 on Build GRCh38
rs777181713 Chr.18: 23753824-23753824 on Build GRCh38
rs367562119 Chr.18: 23753712-23753712 on Build GRCh38
rs147502831 Chr.18: 23753703-23753703 on Build GRCh38
rs62093355 Chr.18: 23753756-23753756 on Build GRCh38
rs544795876 Chr.18: 23753826-23753826 on Build GRCh38
rs779891910 Chr.18: 23753745-23753745 on Build GRCh38
rs997070424 Chr.18: 23753850-23753850 on Build GRCh38
rs771405735 Chr.18: 23753781-23753781 on Build GRCh38
rs1044444092 Chr.18: 23753682-23753682 on Build GRCh38
rs748927008 Chr.18: 23753747-23753747 on Build GRCh38
rs773896979 Chr.18: 23753831-23753831 on Build GRCh38
rs747728113 Chr.18: 23753757-23753757 on Build GRCh38
rs747582703 Chr.18: 23753777-23753777 on Build GRCh38
rs759892468 Chr.18: 23753715-23753715 on Build GRCh38
rs527870822 Chr.18: 23753729-23753729 on Build GRCh38
rs765526931 Chr.18: 23753845-23753845 on Build GRCh38
rs1009665865 Chr.18: 23753876-23753876 on Build GRCh38
rs759643691 Chr.18: 23753841-23753841 on Build GRCh38
rs1019752924 Chr.18: 23753835-23753835 on Build GRCh38
rs565368053 Chr.18: 23753716-23753716 on Build GRCh38
rs1012621879 Chr.18: 23753669-23753669 on Build GRCh38
rs755908558 Chr.18: 23753737-23753737 on Build GRCh38
rs1004725565 Chr.18: 23753759-23753759 on Build GRCh38
rs372217706 Chr.18: 23753717-23753717 on Build GRCh38
rs762207910 Chr.18: 23753685-23753685 on Build GRCh38
rs768048429 Chr.18: 23753675-23753675 on Build GRCh38
rs376609554 Chr.18: 23753711-23753711 on Build GRCh38
rs905474134 Chr.18: 23753686-23753686 on Build GRCh38
rs541234075 Chr.18: 23753748-23753748 on Build GRCh38
rs1054966863 Chr.18: 23753676-23753676 on Build GRCh38
rs772413926 Chr.18: 23753763-23753763 on Build GRCh38
rs757101344 Chr.18: 23753719-23753719 on Build GRCh38
rs562833654 Chr.18: 23753681-23753681 on Build GRCh38
rs754105676 Chr.18: 23753713-23753713 on Build GRCh38

View Full Product Details