Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111776060-111776313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCND3
Entrez Gene ID
3752
Gene Name
potassium voltage-gated channel subfamily D member 3
Gene Aliases
BRGDA9, KCND3L, KCND3S, KSHIVB, KV4.3, SCA19, SCA22
UniGene
Hs.666367
Gene Chromosome Location
Chr.1: 111770662-111989577 on Build GRCh38
Transcripts
NM_004980.4 NM_172198.2 XM_005270851.4 XM_006710629.3 XM_006710630.3 XM_006710631.3 XM_017001244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3471289 c.1799G>A p.G600E Chr.1: 111776246-111776246 on Build GRCh38
4420108 c.1805G>C p.R602T Chr.1: 111776240-111776240 on Build GRCh38
1332732 c.1863G>A p.T621T Chr.1: 111776125-111776125 on Build GRCh38
7305336 c.1737C>T p.D579D Chr.1: 111776251-111776251 on Build GRCh38
4859616 c.1813T>A p.C605S Chr.1: 111776232-111776232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895254433 Chr.1: 111776189-111776189 on Build GRCh38
rs201275860 Chr.1: 111776204-111776204 on Build GRCh38
rs745523741 Chr.1: 111776248-111776248 on Build GRCh38
rs751182609 Chr.1: 111776203-111776203 on Build GRCh38
rs777172603 Chr.1: 111776128-111776128 on Build GRCh38
rs149344567 Chr.1: 111776247-111776247 on Build GRCh38
rs372362132 Chr.1: 111776166-111776166 on Build GRCh38
rs766650482 Chr.1: 111776179-111776179 on Build GRCh38
rs757457679 Chr.1: 111776084-111776085 on Build GRCh38
rs760710842 Chr.1: 111776169-111776169 on Build GRCh38
rs113196892 Chr.1: 111776249-111776249 on Build GRCh38
rs766562520 Chr.1: 111776102-111776102 on Build GRCh38
rs772579682 Chr.1: 111776163-111776163 on Build GRCh38
rs758286812 Chr.1: 111776127-111776127 on Build GRCh38
rs199663609 Chr.1: 111776180-111776180 on Build GRCh38
rs903701604 Chr.1: 111776273-111776273 on Build GRCh38
rs750478656 Chr.1: 111776098-111776098 on Build GRCh38
rs760907112 Chr.1: 111776099-111776099 on Build GRCh38
rs746497556 Chr.1: 111776132-111776132 on Build GRCh38
rs754759010 Chr.1: 111776121-111776121 on Build GRCh38
rs748519692 Chr.1: 111776277-111776277 on Build GRCh38
rs147087785 Chr.1: 111776086-111776086 on Build GRCh38
rs948125814 Chr.1: 111776196-111776196 on Build GRCh38
rs190953266 Chr.1: 111776109-111776109 on Build GRCh38
rs753669886 Chr.1: 111776103-111776103 on Build GRCh38
rs774711788 Chr.1: 111776085-111776085 on Build GRCh38
rs752589615 Chr.1: 111776126-111776126 on Build GRCh38
rs772493153 Chr.1: 111776280-111776280 on Build GRCh38
rs910058506 Chr.1: 111776108-111776108 on Build GRCh38
rs147262533 Chr.1: 111776155-111776155 on Build GRCh38
rs939370048 Chr.1: 111776162-111776162 on Build GRCh38
rs779007324 Chr.1: 111776212-111776212 on Build GRCh38
rs758318696 Chr.1: 111776186-111776186 on Build GRCh38
rs375391961 Chr.1: 111776243-111776243 on Build GRCh38
rs777736918 Chr.1: 111776200-111776200 on Build GRCh38
rs762261334 Chr.1: 111776158-111776158 on Build GRCh38
rs752495973 Chr.1: 111776184-111776184 on Build GRCh38
rs774713377 Chr.1: 111776156-111776156 on Build GRCh38
rs181971539 Chr.1: 111776251-111776251 on Build GRCh38
rs780988439 Chr.1: 111776148-111776148 on Build GRCh38
rs770747636 Chr.1: 111776140-111776140 on Build GRCh38
rs767502190 Chr.1: 111776097-111776097 on Build GRCh38
rs773760801 Chr.1: 111776284-111776284 on Build GRCh38
rs756814713 Chr.1: 111776223-111776223 on Build GRCh38
rs944024237 Chr.1: 111776167-111776167 on Build GRCh38
rs186194682 Chr.1: 111776276-111776276 on Build GRCh38

View Full Product Details