Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49908345-49908604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4I1
Entrez Gene ID
259307
Gene Name
interleukin 4 induced 1
Gene Aliases
FIG1, LAAO, LAO
UniGene
Hs.574492
Gene Chromosome Location
Chr.19: 49889654-49929539 on Build GRCh38
Transcripts
NM_001258017.1 NM_001258018.1 NM_172374.2 NR_047577.1 NM_001193357.1 NM_012346.4 NM_016553.4 NM_153718.3 NM_153719.3

Gene Symbol
NUP62
Entrez Gene ID
23636
Gene Name
nucleoporin 62
Gene Aliases
IBSN, SNDI, p62
UniGene
Hs.574492
Gene Chromosome Location
Chr.19: 49906825-49929731 on Build GRCh38
Transcripts
NM_001258017.1 NM_001258018.1 NM_172374.2 NR_047577.1 NM_001193357.1 NM_012346.4 NM_016553.4 NM_153718.3 NM_153719.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
262489 c.1365C>T p.I455I Chr.19: 49908443-49908443 on Build GRCh38
5022044 c.1393G>C p.A465P Chr.19: 49908415-49908415 on Build GRCh38
5013488 c.1213G>T p.D405Y Chr.19: 49908595-49908595 on Build GRCh38
4080248 c.1380G>A p.T460T Chr.19: 49908428-49908428 on Build GRCh38
6731673 c.1304A>G p.K435R Chr.19: 49908504-49908504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372597263 Chr.19: 49908487-49908487 on Build GRCh38
rs143078249 Chr.19: 49908548-49908548 on Build GRCh38
rs751148161 Chr.19: 49908404-49908404 on Build GRCh38
rs763688589 Chr.19: 49908499-49908499 on Build GRCh38
rs534262181 Chr.19: 49908412-49908412 on Build GRCh38
rs753378692 Chr.19: 49908518-49908518 on Build GRCh38
rs146748194 Chr.19: 49908470-49908470 on Build GRCh38
rs61751953 Chr.19: 49908370-49908370 on Build GRCh38
rs766085373 Chr.19: 49908395-49908395 on Build GRCh38
rs759560737 Chr.19: 49908415-49908415 on Build GRCh38
rs149033608 Chr.19: 49908532-49908532 on Build GRCh38
rs892028 Chr.19: 49908485-49908485 on Build GRCh38
rs770574673 Chr.19: 49908479-49908479 on Build GRCh38
rs780484739 Chr.19: 49908428-49908428 on Build GRCh38
rs777922064 Chr.19: 49908547-49908547 on Build GRCh38
rs1056692605 Chr.19: 49908529-49908529 on Build GRCh38
rs1038745473 Chr.19: 49908563-49908563 on Build GRCh38
rs139913264 Chr.19: 49908372-49908372 on Build GRCh38
rs375741773 Chr.19: 49908550-49908550 on Build GRCh38
rs781239867 Chr.19: 49908448-49908448 on Build GRCh38
rs201085106 Chr.19: 49908430-49908430 on Build GRCh38
rs755458660 Chr.19: 49908531-49908531 on Build GRCh38
rs947153787 Chr.19: 49908524-49908524 on Build GRCh38
rs775526160 Chr.19: 49908573-49908573 on Build GRCh38
rs934276321 Chr.19: 49908474-49908474 on Build GRCh38
rs778559091 Chr.19: 49908552-49908552 on Build GRCh38
rs774107779 Chr.19: 49908414-49908414 on Build GRCh38
rs760298987 Chr.19: 49908488-49908488 on Build GRCh38
rs750766145 Chr.19: 49908422-49908422 on Build GRCh38
rs887412812 Chr.19: 49908549-49908549 on Build GRCh38
rs760388870 Chr.19: 49908421-49908421 on Build GRCh38
rs755550156 Chr.19: 49908442-49908442 on Build GRCh38
rs138767204 Chr.19: 49908572-49908572 on Build GRCh38
rs752321450 Chr.19: 49908417-49908417 on Build GRCh38
rs747579103 Chr.19: 49908369-49908369 on Build GRCh38
rs143412590 Chr.19: 49908425-49908425 on Build GRCh38
rs151075180 Chr.19: 49908367-49908367 on Build GRCh38
rs774201402 Chr.19: 49908484-49908484 on Build GRCh38
rs368893422 Chr.19: 49908429-49908429 on Build GRCh38
rs144871187 Chr.19: 49908521-49908521 on Build GRCh38
rs140379555 Chr.19: 49908471-49908471 on Build GRCh38
rs776699787 Chr.19: 49908381-49908381 on Build GRCh38
rs745705297 Chr.19: 49908554-49908554 on Build GRCh38
rs376266072 Chr.19: 49908413-49908413 on Build GRCh38
rs756100231 Chr.19: 49908545-49908545 on Build GRCh38
rs748368965 Chr.19: 49908386-49908386 on Build GRCh38
rs755999151 Chr.19: 49908553-49908553 on Build GRCh38
rs879939773 Chr.19: 49908571-49908571 on Build GRCh38
rs751983023 Chr.19: 49908530-49908530 on Build GRCh38
rs761537104 Chr.19: 49908520-49908520 on Build GRCh38
rs753234707 Chr.19: 49908541-49908541 on Build GRCh38
rs767067886 Chr.19: 49908416-49908416 on Build GRCh38
rs1062799 Chr.19: 49908517-49908517 on Build GRCh38
rs748278801 Chr.19: 49908469-49908469 on Build GRCh38
rs780463655 Chr.19: 49908368-49908368 on Build GRCh38
rs917797148 Chr.19: 49908533-49908533 on Build GRCh38

View Full Product Details