Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 143712099-143712360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHACTR2
Entrez Gene ID
9749
Gene Name
phosphatase and actin regulator 2
Gene Aliases
C6orf56
UniGene
Hs.102471
Gene Chromosome Location
Chr.6: 143608180-143831185 on Build GRCh38
Transcripts
NM_001100164.1 NM_001100165.1 NM_001100166.1 NM_014721.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1262020 c.205A>T p.T69S Chr.6: 143712174-143712174 on Build GRCh38
1719984 c.112G>T p.G38C Chr.6: 143712114-143712114 on Build GRCh38
5661670 c.175delA p.K60fs*14 Chr.6: 143712144-143712144 on Build GRCh38
6858088 c.173C>G p.T58S Chr.6: 143712175-143712175 on Build GRCh38
1074059 c.190G>A p.D64N Chr.6: 143712159-143712159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199516062 Chr.6: 143712131-143712131 on Build GRCh38
rs939373056 Chr.6: 143712218-143712218 on Build GRCh38
rs564611893 Chr.6: 143712141-143712141 on Build GRCh38
rs781179766 Chr.6: 143712187-143712187 on Build GRCh38
rs897876974 Chr.6: 143712259-143712260 on Build GRCh38
rs372794508 Chr.6: 143712240-143712240 on Build GRCh38
rs993615413 Chr.6: 143712263-143712264 on Build GRCh38
rs3762004 Chr.6: 143712229-143712229 on Build GRCh38
rs1005417289 Chr.6: 143712307-143712311 on Build GRCh38
rs575741318 Chr.6: 143712158-143712158 on Build GRCh38
rs986798973 Chr.6: 143712324-143712324 on Build GRCh38
rs953536440 Chr.6: 143712285-143712285 on Build GRCh38
rs554733689 Chr.6: 143712157-143712157 on Build GRCh38
rs766664422 Chr.6: 143712178-143712178 on Build GRCh38
rs201524736 Chr.6: 143712179-143712179 on Build GRCh38
rs1025391026 Chr.6: 143712264-143712264 on Build GRCh38
rs748227010 Chr.6: 143712228-143712228 on Build GRCh38
rs750884444 Chr.6: 143712143-143712143 on Build GRCh38
rs765767795 Chr.6: 143712135-143712135 on Build GRCh38
rs377171403 Chr.6: 143712289-143712289 on Build GRCh38
rs989701513 Chr.6: 143712204-143712204 on Build GRCh38
rs868610394 Chr.6: 143712159-143712159 on Build GRCh38

View Full Product Details