Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6646671-6646920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1363797 c.208A>G p.N70D Chr.12: 6646848-6646848 on Build GRCh38
4575680 c.146T>C p.L49P Chr.12: 6646910-6646910 on Build GRCh38
7354011 c.151A>T p.T51S Chr.12: 6646905-6646905 on Build GRCh38
694700 c.150G>A p.L50L Chr.12: 6646906-6646906 on Build GRCh38
273851 c.175A>G p.T59A Chr.12: 6646881-6646881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984458145 Chr.12: 6646737-6646737 on Build GRCh38
rs751804265 Chr.12: 6646858-6646858 on Build GRCh38
rs772753142 Chr.12: 6646792-6646792 on Build GRCh38
rs746219680 Chr.12: 6646773-6646773 on Build GRCh38
rs990532603 Chr.12: 6646830-6646830 on Build GRCh38
rs773999362 Chr.12: 6646797-6646797 on Build GRCh38
rs930508082 Chr.12: 6646870-6646870 on Build GRCh38
rs771779784 Chr.12: 6646819-6646819 on Build GRCh38
rs568383765 Chr.12: 6646851-6646851 on Build GRCh38
rs951735970 Chr.12: 6646709-6646709 on Build GRCh38
rs372560335 Chr.12: 6646801-6646801 on Build GRCh38
rs201181204 Chr.12: 6646899-6646899 on Build GRCh38
rs769877349 Chr.12: 6646757-6646757 on Build GRCh38
rs759401934 Chr.12: 6646810-6646810 on Build GRCh38
rs907807595 Chr.12: 6646900-6646900 on Build GRCh38
rs752892968 Chr.12: 6646879-6646879 on Build GRCh38
rs777889689 Chr.12: 6646760-6646760 on Build GRCh38
rs1020565768 Chr.12: 6646713-6646713 on Build GRCh38
rs775291443 Chr.12: 6646827-6646827 on Build GRCh38
rs202009678 Chr.12: 6646777-6646777 on Build GRCh38
rs748160304 Chr.12: 6646745-6646745 on Build GRCh38
rs1034642815 Chr.12: 6646768-6646768 on Build GRCh38
rs145343092 Chr.12: 6646850-6646850 on Build GRCh38
rs927024421 Chr.12: 6646708-6646708 on Build GRCh38
rs760458702 Chr.12: 6646836-6646836 on Build GRCh38
rs766923238 Chr.12: 6646857-6646857 on Build GRCh38
rs763691389 Chr.12: 6646837-6646837 on Build GRCh38
rs777823615 Chr.12: 6646894-6646894 on Build GRCh38
rs960329346 Chr.12: 6646805-6646805 on Build GRCh38
rs979836518 Chr.12: 6646702-6646702 on Build GRCh38
rs749180717 Chr.12: 6646772-6646772 on Build GRCh38
rs200384019 Chr.12: 6646869-6646869 on Build GRCh38
rs968017534 Chr.12: 6646736-6646736 on Build GRCh38
rs141590474 Chr.12: 6646795-6646795 on Build GRCh38
rs756366387 Chr.12: 6646884-6646884 on Build GRCh38

View Full Product Details