Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 44575809-44576034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSC22D1
Entrez Gene ID
8848
Gene Name
TSC22 domain family member 1
Gene Aliases
Ptg-2, TGFB1I4, TSC22
UniGene
Hs.436383
Gene Chromosome Location
Chr.13: 44432143-44576565 on Build GRCh38
Transcripts
NM_001243799.1 NM_183422.3 XM_005266583.4 XM_017020810.1 XM_017020811.1 NR_038381.1

Gene Symbol
TSC22D1-AS1
Entrez Gene ID
641467
Gene Name
TSC22D1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.624196
Gene Chromosome Location
Chr.13: 44575896-44580432 on Build GRCh38
Transcripts
NM_001243799.1 NM_183422.3 XM_005266583.4 XM_017020810.1 XM_017020811.1 NR_038381.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810302 c.201G>A p.P67P Chr.13: 44575874-44575874 on Build GRCh38
7252576 c.158C>T p.T53I Chr.13: 44575917-44575917 on Build GRCh38
432421 c.197C>T p.P66L Chr.13: 44575878-44575878 on Build GRCh38
1268764 c.185C>T p.S62L Chr.13: 44575890-44575890 on Build GRCh38
7294446 c.68C>T p.A23V Chr.13: 44576007-44576007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910783282 Chr.13: 44575865-44575865 on Build GRCh38
rs149004960 Chr.13: 44575999-44575999 on Build GRCh38
rs758412462 Chr.13: 44575860-44575860 on Build GRCh38
rs377521420 Chr.13: 44576005-44576005 on Build GRCh38
rs146669731 Chr.13: 44575988-44575988 on Build GRCh38
rs986444610 Chr.13: 44575846-44575846 on Build GRCh38
rs781611396 Chr.13: 44575871-44575871 on Build GRCh38
rs751871252 Chr.13: 44575834-44575835 on Build GRCh38
rs750196577 Chr.13: 44575851-44575851 on Build GRCh38
rs768013522 Chr.13: 44575883-44575883 on Build GRCh38
rs375662160 Chr.13: 44575874-44575874 on Build GRCh38
rs749708622 Chr.13: 44575967-44575967 on Build GRCh38
rs1003409859 Chr.13: 44575946-44575946 on Build GRCh38
rs200325152 Chr.13: 44575896-44575896 on Build GRCh38
rs972226131 Chr.13: 44575944-44575944 on Build GRCh38
rs781737412 Chr.13: 44575950-44575950 on Build GRCh38
rs766171303 Chr.13: 44575902-44575902 on Build GRCh38
rs370534959 Chr.13: 44575877-44575877 on Build GRCh38
rs769962486 Chr.13: 44575953-44575953 on Build GRCh38
rs149542670 Chr.13: 44575931-44575931 on Build GRCh38
rs373794730 Chr.13: 44575866-44575866 on Build GRCh38
rs767393622 Chr.13: 44575844-44575844 on Build GRCh38
rs910542714 Chr.13: 44576010-44576010 on Build GRCh38
rs140514784 Chr.13: 44575837-44575837 on Build GRCh38
rs1046769064 Chr.13: 44575908-44575908 on Build GRCh38
rs762219265 Chr.13: 44575855-44575855 on Build GRCh38
rs771482488 Chr.13: 44575991-44575991 on Build GRCh38
rs760391914 Chr.13: 44575899-44575899 on Build GRCh38
rs776390949 Chr.13: 44576011-44576011 on Build GRCh38
rs905150382 Chr.13: 44575942-44575942 on Build GRCh38
rs770236458 Chr.13: 44575961-44575961 on Build GRCh38
rs371487971 Chr.13: 44575835-44575835 on Build GRCh38
rs761276255 Chr.13: 44575890-44575890 on Build GRCh38
rs752864488 Chr.13: 44575921-44575921 on Build GRCh38
rs767476950 Chr.13: 44575891-44575891 on Build GRCh38
rs773879451 Chr.13: 44575889-44575889 on Build GRCh38
rs757350310 Chr.13: 44575943-44575943 on Build GRCh38
rs762719188 Chr.13: 44575880-44575880 on Build GRCh38
rs954989156 Chr.13: 44575949-44575949 on Build GRCh38
rs746391369 Chr.13: 44575958-44575958 on Build GRCh38
rs759310348 Chr.13: 44576012-44576012 on Build GRCh38
rs567753994 Chr.13: 44575857-44575857 on Build GRCh38
rs1005196767 Chr.13: 44575914-44575914 on Build GRCh38
rs758675937 Chr.13: 44575927-44575927 on Build GRCh38
rs114920421 Chr.13: 44575989-44575989 on Build GRCh38
rs888116545 Chr.13: 44575912-44575912 on Build GRCh38
rs761839804 Chr.13: 44575839-44575839 on Build GRCh38
rs201283059 Chr.13: 44575964-44575964 on Build GRCh38
rs746233880 Chr.13: 44575873-44575873 on Build GRCh38
rs751650153 Chr.13: 44575932-44575932 on Build GRCh38
rs199563545 Chr.13: 44575986-44575986 on Build GRCh38
rs775350515 Chr.13: 44575834-44575834 on Build GRCh38
rs145475008 Chr.13: 44575869-44575869 on Build GRCh38
rs968469696 Chr.13: 44576003-44576003 on Build GRCh38
rs773072144 Chr.13: 44575996-44575996 on Build GRCh38

View Full Product Details