Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34623410-34623683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID3C
Entrez Gene ID
138715
Gene Name
AT-rich interaction domain 3C
Gene Aliases
-
UniGene
Hs.534549
Gene Chromosome Location
Chr.9: 34621360-34633112 on Build GRCh38
Transcripts
NM_001017363.1 XM_017014283.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3779976 c.667G>A p.E223K Chr.9: 34623623-34623623 on Build GRCh38
5463793 c.842T>C p.L281P Chr.9: 34623448-34623448 on Build GRCh38
3907037 c.813C>A p.S271S Chr.9: 34623477-34623477 on Build GRCh38
1166771 c.737delA p.Q246fs*36 Chr.9: 34623553-34623553 on Build GRCh38
6536759 c.607G>C p.E203Q Chr.9: 34623683-34623683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750287236 Chr.9: 34623476-34623476 on Build GRCh38
rs574056651 Chr.9: 34623519-34623519 on Build GRCh38
rs7018560 Chr.9: 34623448-34623448 on Build GRCh38
rs764865701 Chr.9: 34623662-34623662 on Build GRCh38
rs775677901 Chr.9: 34623458-34623458 on Build GRCh38
rs781641801 Chr.9: 34623496-34623496 on Build GRCh38
rs201996168 Chr.9: 34623618-34623618 on Build GRCh38
rs922574481 Chr.9: 34623500-34623500 on Build GRCh38
rs772206919 Chr.9: 34623542-34623542 on Build GRCh38
rs754590551 Chr.9: 34623480-34623480 on Build GRCh38
rs200520372 Chr.9: 34623531-34623531 on Build GRCh38
rs773273193 Chr.9: 34623544-34623544 on Build GRCh38
rs758095175 Chr.9: 34623583-34623584 on Build GRCh38
rs781557631 Chr.9: 34623434-34623434 on Build GRCh38
rs572894320 Chr.9: 34623452-34623452 on Build GRCh38
rs750934791 Chr.9: 34623597-34623597 on Build GRCh38
rs933991567 Chr.9: 34623517-34623517 on Build GRCh38
rs780419870 Chr.9: 34623600-34623600 on Build GRCh38
rs768875626 Chr.9: 34623532-34623532 on Build GRCh38
rs752201658 Chr.9: 34623487-34623487 on Build GRCh38
rs917838166 Chr.9: 34623608-34623608 on Build GRCh38
rs770157441 Chr.9: 34623525-34623525 on Build GRCh38
rs558534855 Chr.9: 34623474-34623474 on Build GRCh38
rs760546230 Chr.9: 34623477-34623477 on Build GRCh38
rs780329882 Chr.9: 34623529-34623529 on Build GRCh38
rs1044541085 Chr.9: 34623550-34623550 on Build GRCh38
rs755197771 Chr.9: 34623607-34623607 on Build GRCh38
rs150908741 Chr.9: 34623449-34623449 on Build GRCh38
rs761923736 Chr.9: 34623541-34623541 on Build GRCh38
rs776337895 Chr.9: 34623652-34623652 on Build GRCh38
rs369136182 Chr.9: 34623596-34623596 on Build GRCh38
rs752188935 Chr.9: 34623589-34623589 on Build GRCh38
rs201965867 Chr.9: 34623522-34623522 on Build GRCh38
rs774346846 Chr.9: 34623540-34623540 on Build GRCh38
rs756575073 Chr.9: 34623599-34623599 on Build GRCh38
rs757750881 Chr.9: 34623491-34623491 on Build GRCh38
rs113833171 Chr.9: 34623582-34623582 on Build GRCh38
rs905493835 Chr.9: 34623564-34623564 on Build GRCh38
rs763538155 Chr.9: 34623594-34623594 on Build GRCh38
rs746210073 Chr.9: 34623443-34623443 on Build GRCh38
rs375123291 Chr.9: 34623605-34623605 on Build GRCh38
rs766276908 Chr.9: 34623579-34623579 on Build GRCh38
rs760628147 Chr.9: 34623551-34623551 on Build GRCh38
rs902619631 Chr.9: 34623645-34623645 on Build GRCh38
rs759342451 Chr.9: 34623658-34623658 on Build GRCh38
rs544733538 Chr.9: 34623470-34623470 on Build GRCh38
rs779196187 Chr.9: 34623613-34623613 on Build GRCh38
rs746297996 Chr.9: 34623524-34623524 on Build GRCh38
rs1000284556 Chr.9: 34623572-34623572 on Build GRCh38
rs904360277 Chr.9: 34623558-34623558 on Build GRCh38
rs776444205 Chr.9: 34623583-34623583 on Build GRCh38
rs774451984 Chr.9: 34623471-34623471 on Build GRCh38
rs77754952 Chr.9: 34623636-34623636 on Build GRCh38
rs766188876 Chr.9: 34623478-34623478 on Build GRCh38
rs540820273 Chr.9: 34623621-34623621 on Build GRCh38
rs770939727 Chr.9: 34623640-34623640 on Build GRCh38
rs7847086 Chr.9: 34623484-34623484 on Build GRCh38
rs773355028 Chr.9: 34623625-34623625 on Build GRCh38
rs752153221 Chr.9: 34623448-34623470 on Build GRCh38
rs757898870 Chr.9: 34623592-34623592 on Build GRCh38
rs753551072 Chr.9: 34623479-34623479 on Build GRCh38
rs996860675 Chr.9: 34623444-34623444 on Build GRCh38
rs759129977 Chr.9: 34623585-34623585 on Build GRCh38
rs764919484 Chr.9: 34623588-34623588 on Build GRCh38

View Full Product Details