Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74674851-74675124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA4F
Entrez Gene ID
10505
Gene Name
ssemaphorin 4F
Gene Aliases
M-SEMA, PRO2353, S4F, SEMAM, SEMAW, m-Sema-M
UniGene
Hs.25887
Gene Chromosome Location
Chr.2: 74654228-74699013 on Build GRCh38
Transcripts
NM_001271661.1 NM_001271662.1 NM_004263.4 NR_073398.1 NR_073399.1 XM_011532475.2 XM_011532476.2 XM_011532477.2 XM_017003172.1 XM_017003173.1 XM_017003174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3583434 c.1070C>T p.P357L Chr.2: 74674956-74674956 on Build GRCh38
391992 c.1074delC p.F358fs*4 Chr.2: 74674960-74674960 on Build GRCh38
5708322 c.1100G>A p.R367K Chr.2: 74674986-74674986 on Build GRCh38
7425781 c.1031C>T p.A344V Chr.2: 74674917-74674917 on Build GRCh38
4169370 c.1139G>T p.R380I Chr.2: 74675025-74675025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749486094 Chr.2: 74674922-74674922 on Build GRCh38
rs370892939 Chr.2: 74674937-74674937 on Build GRCh38
rs765176082 Chr.2: 74675008-74675008 on Build GRCh38
rs774285893 Chr.2: 74674974-74674974 on Build GRCh38
rs182897833 Chr.2: 74674875-74674875 on Build GRCh38
rs767626075 Chr.2: 74675037-74675037 on Build GRCh38
rs748691898 Chr.2: 74674957-74674957 on Build GRCh38
rs942153027 Chr.2: 74675081-74675081 on Build GRCh38
rs756293769 Chr.2: 74675054-74675054 on Build GRCh38
rs772654729 Chr.2: 74674965-74674965 on Build GRCh38
rs774894692 Chr.2: 74675011-74675012 on Build GRCh38
rs980237622 Chr.2: 74675030-74675030 on Build GRCh38
rs780172465 Chr.2: 74675057-74675057 on Build GRCh38
rs363640 Chr.2: 74675101-74675101 on Build GRCh38
rs766181364 Chr.2: 74674886-74674886 on Build GRCh38
rs771563690 Chr.2: 74674963-74674963 on Build GRCh38
rs1038298428 Chr.2: 74674951-74674951 on Build GRCh38
rs750483746 Chr.2: 74675050-74675050 on Build GRCh38
rs746103016 Chr.2: 74674926-74674926 on Build GRCh38
rs748296656 Chr.2: 74675000-74675000 on Build GRCh38
rs143403780 Chr.2: 74674938-74674938 on Build GRCh38
rs200774522 Chr.2: 74675028-74675028 on Build GRCh38
rs754155106 Chr.2: 74675061-74675061 on Build GRCh38
rs911103703 Chr.2: 74674921-74674921 on Build GRCh38
rs149542267 Chr.2: 74674901-74674901 on Build GRCh38
rs71418744 Chr.2: 74674944-74674944 on Build GRCh38
rs752652679 Chr.2: 74675020-74675020 on Build GRCh38
rs760316820 Chr.2: 74674966-74674966 on Build GRCh38
rs776442238 Chr.2: 74674992-74674992 on Build GRCh38
rs1047245446 Chr.2: 74675038-74675038 on Build GRCh38
rs771420587 Chr.2: 74674896-74674896 on Build GRCh38
rs367738524 Chr.2: 74675076-74675076 on Build GRCh38
rs376302910 Chr.2: 74674898-74674898 on Build GRCh38
rs149043845 Chr.2: 74674946-74674946 on Build GRCh38
rs754892545 Chr.2: 74674892-74674892 on Build GRCh38
rs774862148 Chr.2: 74674956-74674956 on Build GRCh38
rs757393861 Chr.2: 74674914-74674914 on Build GRCh38
rs146041751 Chr.2: 74674895-74674895 on Build GRCh38
rs553405604 Chr.2: 74674995-74674995 on Build GRCh38
rs765948654 Chr.2: 74674989-74674989 on Build GRCh38
rs746171617 Chr.2: 74675070-74675087 on Build GRCh38
rs765294557 Chr.2: 74674897-74674897 on Build GRCh38

View Full Product Details