Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101307982-101308239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF149
Entrez Gene ID
284996
Gene Name
ring finger protein 149
Gene Aliases
DNAPTP2
UniGene
Hs.142074
Gene Chromosome Location
Chr.2: 101271213-101308716 on Build GRCh38
Transcripts
NM_173647.3 XM_005263920.2 XM_005263921.3 XM_011510990.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6153003 c.438_439CA>AT p.T147S Chr.2: 101308150-101308151 on Build GRCh38
6767375 c.454C>A p.H152N Chr.2: 101308135-101308135 on Build GRCh38
4855022 c.454C>T p.H152Y Chr.2: 101308135-101308135 on Build GRCh38
6767373 c.426C>A p.R142R Chr.2: 101308163-101308163 on Build GRCh38
4804452 c.381G>T p.A127A Chr.2: 101308208-101308208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910249811 Chr.2: 101308077-101308077 on Build GRCh38
rs767431543 Chr.2: 101308110-101308110 on Build GRCh38
rs202062114 Chr.2: 101308217-101308217 on Build GRCh38
rs905206894 Chr.2: 101308203-101308203 on Build GRCh38
rs768686558 Chr.2: 101308092-101308092 on Build GRCh38
rs555126262 Chr.2: 101308020-101308020 on Build GRCh38
rs987333305 Chr.2: 101308067-101308067 on Build GRCh38
rs374634123 Chr.2: 101308100-101308100 on Build GRCh38
rs769878414 Chr.2: 101308081-101308081 on Build GRCh38
rs915603527 Chr.2: 101308118-101308118 on Build GRCh38
rs754227681 Chr.2: 101308098-101308098 on Build GRCh38
rs780366080 Chr.2: 101308204-101308204 on Build GRCh38
rs754521580 Chr.2: 101308154-101308154 on Build GRCh38
rs752075640 Chr.2: 101308160-101308160 on Build GRCh38
rs778247775 Chr.2: 101308157-101308157 on Build GRCh38
rs761683589 Chr.2: 101308105-101308105 on Build GRCh38
rs954682036 Chr.2: 101308028-101308028 on Build GRCh38
rs762313310 Chr.2: 101308128-101308128 on Build GRCh38
rs138453807 Chr.2: 101308187-101308187 on Build GRCh38
rs765863528 Chr.2: 101308151-101308151 on Build GRCh38
rs997124140 Chr.2: 101308035-101308035 on Build GRCh38
rs760273047 Chr.2: 101308133-101308133 on Build GRCh38
rs919173317 Chr.2: 101308024-101308024 on Build GRCh38
rs368889934 Chr.2: 101308163-101308163 on Build GRCh38
rs1029831452 Chr.2: 101308040-101308040 on Build GRCh38
rs1052651273 Chr.2: 101308115-101308115 on Build GRCh38
rs538224134 Chr.2: 101308018-101308018 on Build GRCh38
rs866615223 Chr.2: 101308166-101308166 on Build GRCh38
rs757551305 Chr.2: 101308161-101308161 on Build GRCh38
rs781441802 Chr.2: 101308164-101308164 on Build GRCh38
rs955555880 Chr.2: 101308043-101308043 on Build GRCh38
rs963560221 Chr.2: 101308005-101308005 on Build GRCh38
rs964275786 Chr.2: 101308026-101308026 on Build GRCh38
rs908289418 Chr.2: 101308082-101308082 on Build GRCh38
rs528164520 Chr.2: 101308091-101308091 on Build GRCh38
rs534016577 Chr.2: 101308142-101308142 on Build GRCh38
rs749376678 Chr.2: 101308206-101308206 on Build GRCh38
rs568562496 Chr.2: 101308113-101308113 on Build GRCh38
rs143827530 Chr.2: 101308173-101308173 on Build GRCh38

View Full Product Details