Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140639684-140639945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCO6
Entrez Gene ID
55374
Gene Name
transmembrane and coiled-coil domains 6
Gene Aliases
PRO1580
UniGene
Hs.534333
Gene Chromosome Location
Chr.5: 140596529-140647411 on Build GRCh38
Transcripts
NM_001300980.1 NM_001300982.1 NM_018502.4 XM_005268477.1 XM_011537663.2 XM_011537665.2 XM_011537668.2 XM_017009617.1 XM_017009618.1 XM_017009619.1 XM_017009620.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4734148 c.198+1G>A p.? Chr.5: 140639852-140639852 on Build GRCh38
1186848 c.173T>A p.V58E Chr.5: 140639826-140639826 on Build GRCh38
6862692 c.178G>A p.A60T Chr.5: 140639831-140639831 on Build GRCh38
4827003 c.151G>C p.E51Q Chr.5: 140639804-140639804 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750805511 Chr.5: 140639744-140639744 on Build GRCh38
rs748936973 Chr.5: 140639785-140639785 on Build GRCh38
rs558148339 Chr.5: 140639848-140639848 on Build GRCh38
rs996390111 Chr.5: 140639923-140639923 on Build GRCh38
rs771904102 Chr.5: 140639806-140639806 on Build GRCh38
rs777323622 Chr.5: 140639779-140639779 on Build GRCh38
rs760769436 Chr.5: 140639721-140639721 on Build GRCh38
rs765602863 Chr.5: 140639734-140639734 on Build GRCh38
rs757596749 Chr.5: 140639855-140639855 on Build GRCh38
rs112361553 Chr.5: 140639902-140639902 on Build GRCh38
rs775506005 Chr.5: 140639818-140639818 on Build GRCh38
rs780389922 Chr.5: 140639759-140639759 on Build GRCh38
rs765330575 Chr.5: 140639875-140639875 on Build GRCh38
rs1039576939 Chr.5: 140639922-140639922 on Build GRCh38
rs965225089 Chr.5: 140639840-140639840 on Build GRCh38
rs1012237225 Chr.5: 140639789-140639789 on Build GRCh38
rs576404549 Chr.5: 140639885-140639885 on Build GRCh38
rs536708880 Chr.5: 140639839-140639839 on Build GRCh38
rs775599761 Chr.5: 140639718-140639718 on Build GRCh38
rs770536153 Chr.5: 140639798-140639798 on Build GRCh38
rs375544608 Chr.5: 140639752-140639752 on Build GRCh38
rs752065487 Chr.5: 140639768-140639768 on Build GRCh38
rs766744207 Chr.5: 140639894-140639894 on Build GRCh38
rs201864344 Chr.5: 140639724-140639724 on Build GRCh38
rs763165234 Chr.5: 140639889-140639889 on Build GRCh38
rs755564566 Chr.5: 140639777-140639777 on Build GRCh38
rs768848338 Chr.5: 140639846-140639846 on Build GRCh38
rs762074975 Chr.5: 140639852-140639852 on Build GRCh38
rs771854330 Chr.5: 140639714-140639714 on Build GRCh38
rs370143925 Chr.5: 140639725-140639725 on Build GRCh38
rs963587032 Chr.5: 140639720-140639720 on Build GRCh38
rs919253654 Chr.5: 140639741-140639741 on Build GRCh38
rs778649338 Chr.5: 140639800-140639800 on Build GRCh38
rs999166229 Chr.5: 140639722-140639722 on Build GRCh38
rs777853704 Chr.5: 140639804-140639804 on Build GRCh38
rs759493690 Chr.5: 140639707-140639707 on Build GRCh38
rs569736667 Chr.5: 140639726-140639726 on Build GRCh38

View Full Product Details