Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 95181117-95181365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF2
Entrez Gene ID
7549
Gene Name
zinc finger protein 2
Gene Aliases
A1-5, ZNF661, Zfp661
UniGene
Hs.590916
Gene Chromosome Location
Chr.2: 95165417-95184317 on Build GRCh38
Transcripts
NM_001017396.2 NM_001282398.1 NM_001291604.1 NM_001291605.1 NM_021088.3

SNP Detail

SNP ID Chromosome Location
rs751944935 Chr.2: 95181154-95181154 on Build GRCh38
rs371584382 Chr.2: 95181139-95181139 on Build GRCh38
rs755580845 Chr.2: 95181182-95181182 on Build GRCh38
rs369624119 Chr.2: 95181303-95181303 on Build GRCh38
rs756862057 Chr.2: 95181158-95181158 on Build GRCh38
rs397802135 Chr.2: 95181299-95181301 on Build GRCh38
rs759495054 Chr.2: 95181170-95181171 on Build GRCh38
rs375372488 Chr.2: 95181287-95181287 on Build GRCh38
rs866167883 Chr.2: 95181276-95181276 on Build GRCh38
rs35211791 Chr.2: 95181321-95181322 on Build GRCh38
rs754877879 Chr.2: 95181308-95181308 on Build GRCh38
rs367697676 Chr.2: 95181255-95181255 on Build GRCh38
rs749233672 Chr.2: 95181208-95181208 on Build GRCh38
rs764420752 Chr.2: 95181140-95181140 on Build GRCh38
rs374343221 Chr.2: 95181159-95181159 on Build GRCh38
rs750115077 Chr.2: 95181298-95181298 on Build GRCh38
rs758230634 Chr.2: 95181321-95181321 on Build GRCh38
rs774591055 Chr.2: 95181289-95181289 on Build GRCh38
rs777684229 Chr.2: 95181342-95181342 on Build GRCh38
rs760298242 Chr.2: 95181301-95181301 on Build GRCh38
rs112887855 Chr.2: 95181299-95181299 on Build GRCh38
rs762186875 Chr.2: 95181292-95181292 on Build GRCh38
rs778641035 Chr.2: 95181238-95181238 on Build GRCh38
rs775623251 Chr.2: 95181282-95181282 on Build GRCh38
rs752460653 Chr.2: 95181319-95181319 on Build GRCh38
rs930918099 Chr.2: 95181273-95181273 on Build GRCh38
rs549101347 Chr.2: 95181304-95181304 on Build GRCh38
rs80249426 Chr.2: 95181265-95181265 on Build GRCh38
rs771913395 Chr.2: 95181204-95181206 on Build GRCh38
rs754930900 Chr.2: 95181220-95181220 on Build GRCh38
rs774203529 Chr.2: 95181277-95181277 on Build GRCh38
rs774002600 Chr.2: 95181190-95181190 on Build GRCh38
rs546284554 Chr.2: 95181207-95181207 on Build GRCh38
rs371994815 Chr.2: 95181311-95181311 on Build GRCh38
rs760453004 Chr.2: 95181304-95181304 on Build GRCh38
rs767187730 Chr.2: 95181297-95181297 on Build GRCh38
rs771295055 Chr.2: 95181256-95181256 on Build GRCh38
rs776948216 Chr.2: 95181264-95181264 on Build GRCh38
rs779507262 Chr.2: 95181195-95181195 on Build GRCh38
rs749956806 Chr.2: 95181179-95181179 on Build GRCh38
rs182498837 Chr.2: 95181283-95181283 on Build GRCh38
rs868789064 Chr.2: 95181210-95181210 on Build GRCh38

View Full Product Details