Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1946717-1946978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL3L
Entrez Gene ID
6123
Gene Name
ribosomal protein L3 like
Gene Aliases
-
UniGene
Hs.657266
Gene Chromosome Location
Chr.16: 1944571-1955123 on Build GRCh38
Transcripts
NM_005061.2 XM_011522571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4722623 c.830G>A p.R277H Chr.16: 1946957-1946957 on Build GRCh38
3957273 c.849+9T>A p.? Chr.16: 1946929-1946929 on Build GRCh38
3957272 c.849+10C>A p.? Chr.16: 1946928-1946928 on Build GRCh38
2129327 c.811C>T p.Q271* Chr.16: 1946976-1946976 on Build GRCh38
5543405 c.823C>T p.H275Y Chr.16: 1946964-1946964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763473869 Chr.16: 1946908-1946908 on Build GRCh38
rs764557044 Chr.16: 1946739-1946739 on Build GRCh38
rs750439718 Chr.16: 1946756-1946756 on Build GRCh38
rs1032242218 Chr.16: 1946881-1946881 on Build GRCh38
rs575430250 Chr.16: 1946910-1946910 on Build GRCh38
rs377171240 Chr.16: 1946746-1946746 on Build GRCh38
rs376756280 Chr.16: 1946917-1946917 on Build GRCh38
rs969204844 Chr.16: 1946805-1946805 on Build GRCh38
rs764528932 Chr.16: 1946914-1946914 on Build GRCh38
rs557944117 Chr.16: 1946888-1946888 on Build GRCh38
rs1025074919 Chr.16: 1946804-1946804 on Build GRCh38
rs773064384 Chr.16: 1946898-1946898 on Build GRCh38
rs149645742 Chr.16: 1946954-1946954 on Build GRCh38
rs1046516289 Chr.16: 1946941-1946941 on Build GRCh38
rs756256234 Chr.16: 1946757-1946757 on Build GRCh38
rs774847994 Chr.16: 1946808-1946809 on Build GRCh38
rs747919233 Chr.16: 1946891-1946891 on Build GRCh38
rs890647496 Chr.16: 1946873-1946873 on Build GRCh38
rs1007621722 Chr.16: 1946877-1946877 on Build GRCh38
rs144221138 Chr.16: 1946958-1946958 on Build GRCh38
rs915541326 Chr.16: 1946826-1946826 on Build GRCh38
rs138672485 Chr.16: 1946738-1946738 on Build GRCh38
rs757450097 Chr.16: 1946745-1946745 on Build GRCh38
rs199832108 Chr.16: 1946899-1946899 on Build GRCh38
rs758673699 Chr.16: 1946778-1946778 on Build GRCh38
rs991211112 Chr.16: 1946817-1946817 on Build GRCh38
rs897846334 Chr.16: 1946786-1946786 on Build GRCh38
rs201584875 Chr.16: 1946900-1946900 on Build GRCh38
rs112468975 Chr.16: 1946796-1946796 on Build GRCh38
rs751898502 Chr.16: 1946743-1946743 on Build GRCh38
rs750706495 Chr.16: 1946920-1946920 on Build GRCh38
rs201166491 Chr.16: 1946916-1946916 on Build GRCh38
rs919497199 Chr.16: 1946852-1946852 on Build GRCh38
rs958224631 Chr.16: 1946887-1946887 on Build GRCh38
rs200828889 Chr.16: 1946933-1946933 on Build GRCh38
rs775922991 Chr.16: 1946907-1946908 on Build GRCh38
rs753314167 Chr.16: 1946736-1946736 on Build GRCh38
rs113529449 Chr.16: 1946937-1946937 on Build GRCh38
rs199867506 Chr.16: 1946924-1946924 on Build GRCh38
rs372569047 Chr.16: 1946892-1946892 on Build GRCh38
rs535829664 Chr.16: 1946841-1946841 on Build GRCh38
rs888050438 Chr.16: 1946737-1946737 on Build GRCh38
rs373185822 Chr.16: 1946872-1946872 on Build GRCh38
rs553982725 Chr.16: 1946842-1946842 on Build GRCh38
rs778718587 Chr.16: 1946946-1946946 on Build GRCh38
rs376186576 Chr.16: 1946957-1946957 on Build GRCh38
rs139391846 Chr.16: 1946953-1946953 on Build GRCh38
rs754897318 Chr.16: 1946940-1946940 on Build GRCh38
rs766645803 Chr.16: 1946797-1946797 on Build GRCh38
rs538119619 Chr.16: 1946863-1946863 on Build GRCh38
rs180887483 Chr.16: 1946828-1946828 on Build GRCh38
rs547027561 Chr.16: 1946767-1946767 on Build GRCh38
rs200817040 Chr.16: 1946889-1946889 on Build GRCh38
rs569184149 Chr.16: 1946859-1946859 on Build GRCh38
rs775826086 Chr.16: 1946902-1946902 on Build GRCh38
rs200719908 Chr.16: 1946923-1946923 on Build GRCh38
rs778165909 Chr.16: 1946779-1946779 on Build GRCh38
rs959758456 Chr.16: 1946799-1946799 on Build GRCh38

View Full Product Details