Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29694846-29695109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
QPRT
Entrez Gene ID
23475
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
HEL-S-90n, QPRTase
UniGene
Hs.513484
Gene Chromosome Location
Chr.16: 29670588-29698699 on Build GRCh38
Transcripts
NM_001318249.1 NM_001318250.1 NM_014298.4 NR_134536.1 XM_005255223.3 XM_017023101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6448788 c.243G>A p.L81L Chr.16: 29694893-29694893 on Build GRCh38
6555591 c.327C>T p.A109A Chr.16: 29694977-29694977 on Build GRCh38
6760963 c.329G>A p.R110H Chr.16: 29694979-29694979 on Build GRCh38
4847627 c.215C>G p.S72C Chr.16: 29694865-29694865 on Build GRCh38
1749541 c.229G>A p.E77K Chr.16: 29694879-29694879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006917414 Chr.16: 29694977-29694977 on Build GRCh38
rs370213499 Chr.16: 29694931-29694931 on Build GRCh38
rs772960109 Chr.16: 29694907-29694907 on Build GRCh38
rs775682684 Chr.16: 29694954-29694954 on Build GRCh38
rs749719593 Chr.16: 29694929-29694929 on Build GRCh38
rs751200843 Chr.16: 29694922-29694922 on Build GRCh38
rs769605728 Chr.16: 29695049-29695049 on Build GRCh38
rs776288906 Chr.16: 29694968-29694968 on Build GRCh38
rs765773484 Chr.16: 29694975-29694975 on Build GRCh38
rs767028115 Chr.16: 29695061-29695061 on Build GRCh38
rs113148463 Chr.16: 29695058-29695058 on Build GRCh38
rs150521045 Chr.16: 29694961-29694961 on Build GRCh38
rs780619295 Chr.16: 29695084-29695084 on Build GRCh38
rs780964711 Chr.16: 29695004-29695004 on Build GRCh38
rs368435241 Chr.16: 29695028-29695028 on Build GRCh38
rs765678157 Chr.16: 29695073-29695073 on Build GRCh38
rs760789006 Chr.16: 29694955-29694955 on Build GRCh38
rs758829109 Chr.16: 29695083-29695083 on Build GRCh38
rs866783667 Chr.16: 29694909-29694909 on Build GRCh38
rs551562862 Chr.16: 29694925-29694925 on Build GRCh38
rs11538879 Chr.16: 29694912-29694912 on Build GRCh38
rs749864953 Chr.16: 29694915-29694915 on Build GRCh38
rs765221759 Chr.16: 29694899-29694899 on Build GRCh38
rs775599319 Chr.16: 29694879-29694879 on Build GRCh38
rs772757445 Chr.16: 29695022-29695022 on Build GRCh38
rs779577006 Chr.16: 29694934-29694934 on Build GRCh38
rs753302206 Chr.16: 29694947-29694947 on Build GRCh38
rs201291109 Chr.16: 29694972-29694972 on Build GRCh38
rs776920602 Chr.16: 29695031-29695031 on Build GRCh38
rs760480330 Chr.16: 29694883-29694883 on Build GRCh38
rs375580837 Chr.16: 29695008-29695008 on Build GRCh38
rs763767975 Chr.16: 29694887-29694887 on Build GRCh38
rs762406073 Chr.16: 29694973-29694973 on Build GRCh38
rs779629901 Chr.16: 29694921-29694921 on Build GRCh38
rs762852677 Chr.16: 29695056-29695056 on Build GRCh38
rs761597812 Chr.16: 29694971-29694971 on Build GRCh38
rs757852504 Chr.16: 29695026-29695027 on Build GRCh38
rs767791140 Chr.16: 29694878-29694878 on Build GRCh38
rs933786087 Chr.16: 29694970-29694970 on Build GRCh38
rs759100664 Chr.16: 29695060-29695060 on Build GRCh38
rs761544874 Chr.16: 29695039-29695039 on Build GRCh38
rs960918475 Chr.16: 29695020-29695020 on Build GRCh38
rs773729446 Chr.16: 29695059-29695059 on Build GRCh38
rs754429942 Chr.16: 29694977-29695005 on Build GRCh38
rs375362968 Chr.16: 29694938-29694938 on Build GRCh38
rs1044184 Chr.16: 29694933-29694933 on Build GRCh38
rs755873089 Chr.16: 29695070-29695070 on Build GRCh38
rs919913651 Chr.16: 29695035-29695035 on Build GRCh38
rs549437266 Chr.16: 29695013-29695013 on Build GRCh38
rs139462668 Chr.16: 29694979-29694979 on Build GRCh38
rs114534146 Chr.16: 29695050-29695050 on Build GRCh38
rs757915160 Chr.16: 29694918-29694918 on Build GRCh38
rs753718338 Chr.16: 29694893-29694893 on Build GRCh38
rs761729022 Chr.16: 29694898-29694898 on Build GRCh38
rs142650560 Chr.16: 29694944-29694944 on Build GRCh38
rs201663383 Chr.16: 29694978-29694978 on Build GRCh38
rs779987312 Chr.16: 29695023-29695023 on Build GRCh38
rs567632718 Chr.16: 29695019-29695019 on Build GRCh38

View Full Product Details