Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77201172-77201445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260379 c.145-4G>T p.? Chr.9: 77201361-77201361 on Build GRCh38
5027083 c.177T>C p.V59V Chr.9: 77201397-77201397 on Build GRCh38
5027085 c.177T>C p.V59V Chr.9: 77201397-77201397 on Build GRCh38
5027084 c.177T>C p.V59V Chr.9: 77201397-77201397 on Build GRCh38
6260377 c.145-4G>T p.? Chr.9: 77201361-77201361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764960108 Chr.9: 77201406-77201406 on Build GRCh38
rs770094234 Chr.9: 77201326-77201326 on Build GRCh38
rs374994178 Chr.9: 77201338-77201338 on Build GRCh38
rs1023712870 Chr.9: 77201348-77201348 on Build GRCh38
rs971049211 Chr.9: 77201408-77201408 on Build GRCh38
rs201490407 Chr.9: 77201392-77201392 on Build GRCh38
rs145306631 Chr.9: 77201386-77201386 on Build GRCh38
rs773748695 Chr.9: 77201342-77201342 on Build GRCh38
rs1057515645 Chr.9: 77201379-77201379 on Build GRCh38
rs373944070 Chr.9: 77201323-77201323 on Build GRCh38
rs750344038 Chr.9: 77201420-77201420 on Build GRCh38
rs899027958 Chr.9: 77201306-77201306 on Build GRCh38
rs958465700 Chr.9: 77201250-77201250 on Build GRCh38
rs150034322 Chr.9: 77201370-77201370 on Build GRCh38
rs754661980 Chr.9: 77201346-77201346 on Build GRCh38
rs970746451 Chr.9: 77201369-77201369 on Build GRCh38
rs1005179599 Chr.9: 77201275-77201275 on Build GRCh38
rs1012645934 Chr.9: 77201298-77201298 on Build GRCh38
rs780764326 Chr.9: 77201318-77201318 on Build GRCh38
rs1037559103 Chr.9: 77201302-77201302 on Build GRCh38
rs759700958 Chr.9: 77201376-77201376 on Build GRCh38
rs1003553884 Chr.9: 77201377-77201377 on Build GRCh38
rs767771206 Chr.9: 77201380-77201380 on Build GRCh38
rs1012023425 Chr.9: 77201344-77201344 on Build GRCh38
rs373128010 Chr.9: 77201209-77201209 on Build GRCh38
rs573008708 Chr.9: 77201361-77201361 on Build GRCh38
rs751415681 Chr.9: 77201353-77201353 on Build GRCh38

View Full Product Details