Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129022325-129022568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3GLB2
Entrez Gene ID
56904
Gene Name
SH3 domain containing GRB2 like endophilin B2
Gene Aliases
PP6569, PP9455, RRIG1
UniGene
Hs.460238
Gene Chromosome Location
Chr.9: 129007792-129028353 on Build GRCh38
Transcripts
NM_001287045.1 NM_001287046.1 NM_020145.3 XM_005252098.2 XM_005252100.2 XM_005252101.3 XM_006717188.2 XM_006717189.2 XM_006717190.1 XM_006717191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105804 c.134G>A p.R45Q Chr.9: 129022353-129022353 on Build GRCh38
6777238 c.121A>C p.N41H Chr.9: 129022366-129022366 on Build GRCh38
6777239 c.121A>C p.N41H Chr.9: 129022366-129022366 on Build GRCh38
4971673 c.144C>T p.S48S Chr.9: 129022343-129022343 on Build GRCh38
3847811 c.150G>T p.K50N Chr.9: 129022337-129022337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372873714 Chr.9: 129022452-129022452 on Build GRCh38
rs531462701 Chr.9: 129022446-129022446 on Build GRCh38
rs774874088 Chr.9: 129022354-129022354 on Build GRCh38
rs773017096 Chr.9: 129022480-129022480 on Build GRCh38
rs17485737 Chr.9: 129022477-129022477 on Build GRCh38
rs751596186 Chr.9: 129022418-129022418 on Build GRCh38
rs749330207 Chr.9: 129022435-129022435 on Build GRCh38
rs138289691 Chr.9: 129022353-129022353 on Build GRCh38
rs968922221 Chr.9: 129022516-129022516 on Build GRCh38
rs559217247 Chr.9: 129022447-129022447 on Build GRCh38
rs780325074 Chr.9: 129022429-129022429 on Build GRCh38
rs142474605 Chr.9: 129022357-129022357 on Build GRCh38
rs906706887 Chr.9: 129022382-129022382 on Build GRCh38
rs376058963 Chr.9: 129022378-129022378 on Build GRCh38
rs749381145 Chr.9: 129022352-129022352 on Build GRCh38
rs747427588 Chr.9: 129022470-129022471 on Build GRCh38
rs899680092 Chr.9: 129022448-129022448 on Build GRCh38
rs761227250 Chr.9: 129022403-129022403 on Build GRCh38
rs916138378 Chr.9: 129022515-129022515 on Build GRCh38
rs750640354 Chr.9: 129022427-129022427 on Build GRCh38
rs377261714 Chr.9: 129022456-129022456 on Build GRCh38
rs79974362 Chr.9: 129022405-129022405 on Build GRCh38
rs772481716 Chr.9: 129022455-129022455 on Build GRCh38
rs780911505 Chr.9: 129022420-129022420 on Build GRCh38
rs991716608 Chr.9: 129022508-129022508 on Build GRCh38
rs996088374 Chr.9: 129022541-129022541 on Build GRCh38
rs968592045 Chr.9: 129022488-129022488 on Build GRCh38
rs759178745 Chr.9: 129022373-129022373 on Build GRCh38
rs764072262 Chr.9: 129022413-129022413 on Build GRCh38
rs756230846 Chr.9: 129022428-129022428 on Build GRCh38
rs762119648 Chr.9: 129022355-129022355 on Build GRCh38
rs775303312 Chr.9: 129022458-129022458 on Build GRCh38
rs752198579 Chr.9: 129022397-129022397 on Build GRCh38
rs778396750 Chr.9: 129022467-129022468 on Build GRCh38
rs762582075 Chr.9: 129022404-129022404 on Build GRCh38

View Full Product Details