Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47922881-47923134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SP2
Entrez Gene ID
6668
Gene Name
Sp2 transcription factor
Gene Aliases
-
UniGene
Hs.514276
Gene Chromosome Location
Chr.17: 47896032-47932271 on Build GRCh38
Transcripts
NM_003110.5 XM_006722023.2 XM_006722025.2 XM_006722026.2 XM_006722027.2 XM_011525136.2 XM_011525137.2 XM_011525138.2 XM_011525139.2 XM_011525140.2 XM_011525142.1 XM_011525143.1 XM_017024968.1 XM_017024969.1 NR_103856.1 NR_103857.1

Gene Symbol
SP2-AS1
Entrez Gene ID
100506325
Gene Name
SP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.656008
Gene Chromosome Location
Chr.17: 47898386-47941404 on Build GRCh38
Transcripts
NM_003110.5 XM_006722023.2 XM_006722025.2 XM_006722026.2 XM_006722027.2 XM_011525136.2 XM_011525137.2 XM_011525138.2 XM_011525139.2 XM_011525140.2 XM_011525142.1 XM_011525143.1 XM_017024968.1 XM_017024969.1 NR_103856.1 NR_103857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1383968 c.1219C>T p.R407* Chr.17: 47923121-47923121 on Build GRCh38
4130336 c.1138T>C p.S380P Chr.17: 47923040-47923040 on Build GRCh38
4651310 c.1133C>T p.A378V Chr.17: 47923035-47923035 on Build GRCh38
5722636 c.1116C>T p.P372P Chr.17: 47923018-47923018 on Build GRCh38
384910 c.1229G>T p.R410L Chr.17: 47923131-47923131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373926962 Chr.17: 47923092-47923092 on Build GRCh38
rs114228308 Chr.17: 47923046-47923046 on Build GRCh38
rs773602999 Chr.17: 47922997-47922997 on Build GRCh38
rs200461216 Chr.17: 47923071-47923071 on Build GRCh38
rs759875168 Chr.17: 47923027-47923027 on Build GRCh38
rs141918529 Chr.17: 47922979-47922979 on Build GRCh38
rs140692820 Chr.17: 47923016-47923016 on Build GRCh38
rs750913732 Chr.17: 47922938-47922938 on Build GRCh38
rs768070728 Chr.17: 47923109-47923109 on Build GRCh38
rs748910049 Chr.17: 47923105-47923105 on Build GRCh38
rs751474678 Chr.17: 47923052-47923052 on Build GRCh38
rs373795992 Chr.17: 47923078-47923078 on Build GRCh38
rs760950338 Chr.17: 47922917-47922917 on Build GRCh38
rs754077754 Chr.17: 47922932-47922932 on Build GRCh38
rs934534927 Chr.17: 47922977-47922977 on Build GRCh38
rs758493874 Chr.17: 47922972-47922972 on Build GRCh38
rs768986864 Chr.17: 47922916-47922916 on Build GRCh38
rs755148313 Chr.17: 47922945-47922945 on Build GRCh38
rs762435181 Chr.17: 47922988-47922988 on Build GRCh38
rs559551268 Chr.17: 47922906-47922906 on Build GRCh38
rs533525682 Chr.17: 47923070-47923070 on Build GRCh38
rs775266914 Chr.17: 47922984-47922984 on Build GRCh38
rs763564375 Chr.17: 47922937-47922938 on Build GRCh38
rs766756737 Chr.17: 47923012-47923012 on Build GRCh38
rs781252649 Chr.17: 47923066-47923066 on Build GRCh38
rs550619098 Chr.17: 47923043-47923043 on Build GRCh38
rs139815805 Chr.17: 47922976-47922976 on Build GRCh38
rs62067380 Chr.17: 47923040-47923040 on Build GRCh38
rs373140962 Chr.17: 47922975-47922975 on Build GRCh38
rs146862346 Chr.17: 47922982-47922982 on Build GRCh38
rs1023082759 Chr.17: 47923096-47923096 on Build GRCh38
rs763441518 Chr.17: 47923045-47923047 on Build GRCh38
rs750935419 Chr.17: 47922911-47922911 on Build GRCh38
rs769619121 Chr.17: 47922983-47922983 on Build GRCh38
rs898078625 Chr.17: 47923047-47923047 on Build GRCh38
rs201442835 Chr.17: 47922922-47922922 on Build GRCh38
rs781086369 Chr.17: 47922980-47922980 on Build GRCh38
rs374059580 Chr.17: 47922951-47922951 on Build GRCh38
rs765504235 Chr.17: 47923039-47923039 on Build GRCh38
rs761191826 Chr.17: 47923003-47923003 on Build GRCh38
rs202014692 Chr.17: 47923053-47923053 on Build GRCh38
rs889426703 Chr.17: 47923041-47923041 on Build GRCh38
rs779518399 Chr.17: 47923017-47923017 on Build GRCh38
rs377738433 Chr.17: 47922955-47922955 on Build GRCh38

View Full Product Details