Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 31934651-31934924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTLL9
Entrez Gene ID
164395
Gene Name
tubulin tyrosine ligase like 9
Gene Aliases
C20orf125
UniGene
Hs.712915
Gene Chromosome Location
Chr.20: 31870702-31944959 on Build GRCh38
Transcripts
NM_001008409.2 NR_134519.1 XM_005260309.2 XM_006723723.2 XM_006723724.2 XM_011528617.1 XM_011528618.1 XM_011528619.1 XM_011528620.2 XM_011528621.1 XM_011528622.1 XM_011528623.2 XM_011528624.1 XM_011528625.1 XM_011528626.1 XM_011528627.2 XM_011528628.1 XM_011528629.1 XM_011528630.1 XM_011528631.1 XM_011528632.1 XM_011528633.1 XM_011528634.1 XM_011528635.1 XM_011528638.1 XM_011528639.1 XM_011528640.1 XM_011528641.1 XM_011528644.1 XM_011528646.1 XM_017027694.1 XM_017027695.1 XM_017027696.1 XM_017027697.1 XM_017027698.1 XM_017027699.1 XM_017027700.1 XM_017027701.1 XM_017027702.1 XM_017027703.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6811619 c.725G>A p.R242H Chr.20: 31934714-31934714 on Build GRCh38
4610206 c.647C>T p.T216M Chr.20: 31934705-31934705 on Build GRCh38
6235042 c.634-2A>T p.? Chr.20: 31934690-31934690 on Build GRCh38
385791 c.816G>A p.A272A Chr.20: 31934733-31934733 on Build GRCh38
4737361 c.762G>T p.E254D Chr.20: 31934751-31934751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750794913 Chr.20: 31934871-31934871 on Build GRCh38
rs779071308 Chr.20: 31934888-31934888 on Build GRCh38
rs200691468 Chr.20: 31934705-31934705 on Build GRCh38
rs367894767 Chr.20: 31934689-31934689 on Build GRCh38
rs758683827 Chr.20: 31934897-31934897 on Build GRCh38
rs963813559 Chr.20: 31934900-31934900 on Build GRCh38
rs776045086 Chr.20: 31934853-31934853 on Build GRCh38
rs753439953 Chr.20: 31934685-31934685 on Build GRCh38
rs370215211 Chr.20: 31934740-31934740 on Build GRCh38
rs746788327 Chr.20: 31934903-31934903 on Build GRCh38
rs745883003 Chr.20: 31934672-31934702 on Build GRCh38
rs757802573 Chr.20: 31934695-31934695 on Build GRCh38
rs370540188 Chr.20: 31934779-31934779 on Build GRCh38
rs774271075 Chr.20: 31934672-31934672 on Build GRCh38
rs201317647 Chr.20: 31934845-31934845 on Build GRCh38
rs566844956 Chr.20: 31934798-31934798 on Build GRCh38
rs766884478 Chr.20: 31934872-31934872 on Build GRCh38
rs568036214 Chr.20: 31934686-31934686 on Build GRCh38
rs761906219 Chr.20: 31934674-31934674 on Build GRCh38
rs373834407 Chr.20: 31934811-31934811 on Build GRCh38
rs780122956 Chr.20: 31934710-31934710 on Build GRCh38
rs557797446 Chr.20: 31934760-31934760 on Build GRCh38
rs767220808 Chr.20: 31934778-31934778 on Build GRCh38
rs771947364 Chr.20: 31934720-31934720 on Build GRCh38
rs760129742 Chr.20: 31934679-31934679 on Build GRCh38
rs751970916 Chr.20: 31934748-31934748 on Build GRCh38
rs775739828 Chr.20: 31934804-31934804 on Build GRCh38
rs749995243 Chr.20: 31934675-31934675 on Build GRCh38
rs533904735 Chr.20: 31934899-31934899 on Build GRCh38
rs375096914 Chr.20: 31934706-31934706 on Build GRCh38
rs755143090 Chr.20: 31934883-31934883 on Build GRCh38
rs748343321 Chr.20: 31934800-31934800 on Build GRCh38
rs377592631 Chr.20: 31934854-31934854 on Build GRCh38
rs148917180 Chr.20: 31934742-31934742 on Build GRCh38
rs747151614 Chr.20: 31934722-31934722 on Build GRCh38
rs767828734 Chr.20: 31934868-31934868 on Build GRCh38
rs370725969 Chr.20: 31934856-31934856 on Build GRCh38
rs377406162 Chr.20: 31934776-31934776 on Build GRCh38
rs369485862 Chr.20: 31934713-31934713 on Build GRCh38
rs779297848 Chr.20: 31934783-31934783 on Build GRCh38
rs764840405 Chr.20: 31934743-31934743 on Build GRCh38
rs373712171 Chr.20: 31934714-31934714 on Build GRCh38
rs761675329 Chr.20: 31934719-31934719 on Build GRCh38
rs767801097 Chr.20: 31934750-31934750 on Build GRCh38
rs773056812 Chr.20: 31934732-31934732 on Build GRCh38
rs773006116 Chr.20: 31934821-31934821 on Build GRCh38
rs751919570 Chr.20: 31934694-31934694 on Build GRCh38
rs754344935 Chr.20: 31934879-31934879 on Build GRCh38
rs772000244 Chr.20: 31934809-31934809 on Build GRCh38
rs199882087 Chr.20: 31934682-31934682 on Build GRCh38
rs775127551 Chr.20: 31934858-31934858 on Build GRCh38
rs758744775 Chr.20: 31934761-31934761 on Build GRCh38
rs757649275 Chr.20: 31934749-31934749 on Build GRCh38
rs748398612 Chr.20: 31934889-31934889 on Build GRCh38
rs17093694 Chr.20: 31934733-31934733 on Build GRCh38

View Full Product Details