Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75649261-75649486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX33
Entrez Gene ID
257364
Gene Name
sorting nexin 33
Gene Aliases
SH3PX3, SH3PXD3C, SNX30
UniGene
Hs.8705
Gene Chromosome Location
Chr.15: 75649007-75658627 on Build GRCh38
Transcripts
NM_001318146.1 NM_153271.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5929611 c.233C>T p.P78L Chr.15: 75649335-75649335 on Build GRCh38
1161139 c.177C>T p.V59V Chr.15: 75649279-75649279 on Build GRCh38
7149630 c.328G>C p.E110Q Chr.15: 75649430-75649430 on Build GRCh38
5441595 c.214A>C p.S72R Chr.15: 75649316-75649316 on Build GRCh38
7133171 c.260G>A p.S87N Chr.15: 75649362-75649362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752159207 Chr.15: 75649363-75649363 on Build GRCh38
rs758262508 Chr.15: 75649301-75649301 on Build GRCh38
rs541973865 Chr.15: 75649343-75649343 on Build GRCh38
rs376935129 Chr.15: 75649310-75649310 on Build GRCh38
rs757022070 Chr.15: 75649304-75649304 on Build GRCh38
rs921019626 Chr.15: 75649422-75649422 on Build GRCh38
rs762631861 Chr.15: 75649461-75649461 on Build GRCh38
rs777717282 Chr.15: 75649302-75649302 on Build GRCh38
rs748655716 Chr.15: 75649408-75649408 on Build GRCh38
rs769238510 Chr.15: 75649329-75649329 on Build GRCh38
rs953788186 Chr.15: 75649433-75649433 on Build GRCh38
rs375635568 Chr.15: 75649375-75649375 on Build GRCh38
rs143460688 Chr.15: 75649468-75649468 on Build GRCh38
rs370275020 Chr.15: 75649465-75649465 on Build GRCh38
rs767329867 Chr.15: 75649370-75649370 on Build GRCh38
rs762452142 Chr.15: 75649332-75649332 on Build GRCh38
rs772474562 Chr.15: 75649435-75649435 on Build GRCh38
rs773510877 Chr.15: 75649346-75649346 on Build GRCh38
rs751304006 Chr.15: 75649303-75649303 on Build GRCh38
rs148674276 Chr.15: 75649359-75649359 on Build GRCh38
rs376433115 Chr.15: 75649446-75649446 on Build GRCh38
rs763640094 Chr.15: 75649366-75649366 on Build GRCh38
rs775099131 Chr.15: 75649454-75649454 on Build GRCh38
rs752158302 Chr.15: 75649284-75649284 on Build GRCh38
rs759174387 Chr.15: 75649355-75649355 on Build GRCh38
rs779203982 Chr.15: 75649401-75649401 on Build GRCh38
rs369471317 Chr.15: 75649379-75649379 on Build GRCh38
rs373531151 Chr.15: 75649442-75649442 on Build GRCh38
rs376544301 Chr.15: 75649400-75649400 on Build GRCh38
rs924301562 Chr.15: 75649409-75649409 on Build GRCh38
rs749572563 Chr.15: 75649397-75649397 on Build GRCh38
rs201133101 Chr.15: 75649307-75649307 on Build GRCh38
rs774979843 Chr.15: 75649331-75649331 on Build GRCh38
rs766961204 Chr.15: 75649337-75649337 on Build GRCh38
rs199900886 Chr.15: 75649441-75649441 on Build GRCh38
rs756320313 Chr.15: 75649376-75649376 on Build GRCh38
rs749411298 Chr.15: 75649324-75649324 on Build GRCh38
rs757207380 Chr.15: 75649368-75649368 on Build GRCh38
rs371112191 Chr.15: 75649438-75649438 on Build GRCh38
rs773908381 Chr.15: 75649466-75649466 on Build GRCh38
rs751465424 Chr.15: 75649367-75649367 on Build GRCh38
rs780114828 Chr.15: 75649319-75649319 on Build GRCh38

View Full Product Details