Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57417879-57418116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF17
Entrez Gene ID
7565
Gene Name
zinc finger protein 17
Gene Aliases
HPF3, KOX10
UniGene
Hs.729403
Gene Chromosome Location
Chr.19: 57409252-57421939 on Build GRCh38
Transcripts
NM_006959.2 XM_017027205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5792401 c.51C>G p.F17L Chr.19: 57417947-57417947 on Build GRCh38
4983966 c.124G>T p.A42S Chr.19: 57418020-57418020 on Build GRCh38
7117774 c.16G>C p.D6H Chr.19: 57417912-57417912 on Build GRCh38
3960608 c.47A>T p.H16L Chr.19: 57417943-57417943 on Build GRCh38
4082057 c.92A>G p.H31R Chr.19: 57417988-57417988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749446523 Chr.19: 57417934-57417934 on Build GRCh38
rs746892285 Chr.19: 57418031-57418031 on Build GRCh38
rs776430933 Chr.19: 57417959-57417959 on Build GRCh38
rs201860126 Chr.19: 57417912-57417912 on Build GRCh38
rs150351433 Chr.19: 57418010-57418010 on Build GRCh38
rs137976299 Chr.19: 57418057-57418057 on Build GRCh38
rs756192587 Chr.19: 57417995-57417995 on Build GRCh38
rs775282945 Chr.19: 57417973-57417973 on Build GRCh38
rs768135558 Chr.19: 57417989-57417989 on Build GRCh38
rs763417537 Chr.19: 57417980-57417980 on Build GRCh38
rs770945133 Chr.19: 57417950-57417950 on Build GRCh38
rs956818450 Chr.19: 57418056-57418056 on Build GRCh38
rs1055093042 Chr.19: 57418021-57418021 on Build GRCh38
rs756176072 Chr.19: 57417932-57417932 on Build GRCh38
rs191501099 Chr.19: 57417998-57417998 on Build GRCh38
rs905750583 Chr.19: 57417999-57417999 on Build GRCh38
rs777579649 Chr.19: 57417939-57417939 on Build GRCh38
rs200011699 Chr.19: 57418039-57418039 on Build GRCh38
rs758221683 Chr.19: 57417936-57417936 on Build GRCh38
rs757606140 Chr.19: 57417961-57417961 on Build GRCh38
rs769367463 Chr.19: 57417971-57417971 on Build GRCh38
rs746762562 Chr.19: 57417940-57417940 on Build GRCh38
rs187048451 Chr.19: 57417909-57417909 on Build GRCh38
rs769455374 Chr.19: 57418052-57418052 on Build GRCh38
rs201137447 Chr.19: 57418002-57418002 on Build GRCh38
rs781474348 Chr.19: 57417993-57417994 on Build GRCh38
rs755246407 Chr.19: 57418006-57418006 on Build GRCh38
rs767887002 Chr.19: 57417914-57417914 on Build GRCh38
rs764388181 Chr.19: 57417978-57417978 on Build GRCh38
rs745729926 Chr.19: 57418051-57418051 on Build GRCh38
rs768393388 Chr.19: 57418079-57418079 on Build GRCh38
rs374334357 Chr.19: 57417933-57417933 on Build GRCh38
rs759892305 Chr.19: 57418068-57418068 on Build GRCh38
rs756992638 Chr.19: 57418033-57418033 on Build GRCh38
rs985111998 Chr.19: 57417918-57417918 on Build GRCh38
rs750554355 Chr.19: 57417927-57417927 on Build GRCh38
rs202180997 Chr.19: 57418058-57418058 on Build GRCh38
rs571027515 Chr.19: 57418030-57418030 on Build GRCh38
rs745425971 Chr.19: 57417961-57417961 on Build GRCh38
rs749131322 Chr.19: 57417952-57417954 on Build GRCh38
rs766604599 Chr.19: 57417977-57417977 on Build GRCh38
rs762050947 Chr.19: 57417986-57417986 on Build GRCh38

View Full Product Details