Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39416702-39416969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAB1
Entrez Gene ID
10454
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 1
Gene Aliases
3'-Tab1, MAP3K7IP1
UniGene
Hs.507681
Gene Chromosome Location
Chr.22: 39399754-39437127 on Build GRCh38
Transcripts
NM_006116.2 NM_153497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1228373 c.367G>A p.D123N Chr.22: 39416833-39416833 on Build GRCh38
3151980 c.370G>A p.A124T Chr.22: 39416836-39416836 on Build GRCh38
4490267 c.360C>T p.S120S Chr.22: 39416826-39416826 on Build GRCh38
7136638 c.366C>T p.D122D Chr.22: 39416832-39416832 on Build GRCh38
4731834 c.331G>A p.D111N Chr.22: 39416797-39416797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777486788 Chr.22: 39416858-39416858 on Build GRCh38
rs118131187 Chr.22: 39416786-39416786 on Build GRCh38
rs377578530 Chr.22: 39416897-39416897 on Build GRCh38
rs373034533 Chr.22: 39416892-39416892 on Build GRCh38
rs747193223 Chr.22: 39416884-39416884 on Build GRCh38
rs28481724 Chr.22: 39416796-39416796 on Build GRCh38
rs765876191 Chr.22: 39416820-39416820 on Build GRCh38
rs367668744 Chr.22: 39416745-39416745 on Build GRCh38
rs376706024 Chr.22: 39416837-39416837 on Build GRCh38
rs987747573 Chr.22: 39416726-39416726 on Build GRCh38
rs755997019 Chr.22: 39416750-39416750 on Build GRCh38
rs528153263 Chr.22: 39416882-39416882 on Build GRCh38
rs367926859 Chr.22: 39416774-39416774 on Build GRCh38
rs567476522 Chr.22: 39416865-39416865 on Build GRCh38
rs915757027 Chr.22: 39416939-39416939 on Build GRCh38
rs202132585 Chr.22: 39416835-39416835 on Build GRCh38
rs749107581 Chr.22: 39416862-39416862 on Build GRCh38
rs546710697 Chr.22: 39416906-39416906 on Build GRCh38
rs758966357 Chr.22: 39416905-39416905 on Build GRCh38
rs368808459 Chr.22: 39416874-39416874 on Build GRCh38
rs777568571 Chr.22: 39416752-39416752 on Build GRCh38
rs371761512 Chr.22: 39416747-39416747 on Build GRCh38
rs545312200 Chr.22: 39416765-39416765 on Build GRCh38
rs760329335 Chr.22: 39416918-39416918 on Build GRCh38
rs749158510 Chr.22: 39416756-39416756 on Build GRCh38
rs373883129 Chr.22: 39416836-39416836 on Build GRCh38
rs752162547 Chr.22: 39416915-39416915 on Build GRCh38
rs913502457 Chr.22: 39416729-39416729 on Build GRCh38
rs777144882 Chr.22: 39416784-39416784 on Build GRCh38
rs774369870 Chr.22: 39416763-39416763 on Build GRCh38
rs376402719 Chr.22: 39416781-39416781 on Build GRCh38
rs201560638 Chr.22: 39416893-39416893 on Build GRCh38
rs770872506 Chr.22: 39416757-39416757 on Build GRCh38
rs778865110 Chr.22: 39416873-39416873 on Build GRCh38
rs754564216 Chr.22: 39416832-39416832 on Build GRCh38
rs762306737 Chr.22: 39416896-39416896 on Build GRCh38
rs12165632 Chr.22: 39416737-39416737 on Build GRCh38
rs763771000 Chr.22: 39416922-39416922 on Build GRCh38
rs967654493 Chr.22: 39416885-39416885 on Build GRCh38
rs760940405 Chr.22: 39416779-39416779 on Build GRCh38
rs772257620 Chr.22: 39416875-39416875 on Build GRCh38
rs1036335172 Chr.22: 39416900-39416900 on Build GRCh38
rs548863608 Chr.22: 39416838-39416838 on Build GRCh38
rs370649149 Chr.22: 39416928-39416928 on Build GRCh38
rs751080763 Chr.22: 39416827-39416827 on Build GRCh38
rs1050602401 Chr.22: 39416857-39416857 on Build GRCh38
rs34571268 Chr.22: 39416764-39416764 on Build GRCh38

View Full Product Details