Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112656893-112657164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC20A1
Entrez Gene ID
6574
Gene Name
solute carrier family 20 member 1
Gene Aliases
GLVR1, Glvr-1, PIT1, PiT-1
UniGene
Hs.187946
Gene Chromosome Location
Chr.2: 112645857-112663825 on Build GRCh38
Transcripts
NM_005415.4 XM_017004768.1 XM_017004769.1 XM_017004770.1 XM_017004771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6267531 c.665G>T p.G222V Chr.2: 112657128-112657128 on Build GRCh38
5539063 c.679C>T p.P227S Chr.2: 112657142-112657142 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751667022 Chr.2: 112657071-112657071 on Build GRCh38
rs749693461 Chr.2: 112657011-112657012 on Build GRCh38
rs190112076 Chr.2: 112656945-112656945 on Build GRCh38
rs756744877 Chr.2: 112657017-112657017 on Build GRCh38
rs371055700 Chr.2: 112657092-112657092 on Build GRCh38
rs771109654 Chr.2: 112657136-112657137 on Build GRCh38
rs200081005 Chr.2: 112657025-112657025 on Build GRCh38
rs767761697 Chr.2: 112657076-112657076 on Build GRCh38
rs768224568 Chr.2: 112656952-112656952 on Build GRCh38
rs72823508 Chr.2: 112657021-112657021 on Build GRCh38
rs562748607 Chr.2: 112657081-112657081 on Build GRCh38
rs756656933 Chr.2: 112657085-112657085 on Build GRCh38
rs114312260 Chr.2: 112656997-112656997 on Build GRCh38
rs116413027 Chr.2: 112656931-112656931 on Build GRCh38
rs768266967 Chr.2: 112657042-112657042 on Build GRCh38
rs778472490 Chr.2: 112657091-112657091 on Build GRCh38
rs947313696 Chr.2: 112656950-112656950 on Build GRCh38
rs72948485 Chr.2: 112656961-112656961 on Build GRCh38
rs766512297 Chr.2: 112657069-112657069 on Build GRCh38
rs761737269 Chr.2: 112657056-112657056 on Build GRCh38
rs1052978537 Chr.2: 112656936-112656936 on Build GRCh38
rs781125202 Chr.2: 112656916-112656916 on Build GRCh38
rs549126171 Chr.2: 112657039-112657039 on Build GRCh38
rs1029237732 Chr.2: 112657116-112657117 on Build GRCh38
rs765201259 Chr.2: 112657062-112657062 on Build GRCh38
rs773258069 Chr.2: 112657064-112657064 on Build GRCh38
rs762975605 Chr.2: 112657066-112657066 on Build GRCh38
rs553199630 Chr.2: 112657011-112657011 on Build GRCh38
rs367664642 Chr.2: 112657140-112657140 on Build GRCh38
rs774955240 Chr.2: 112657038-112657038 on Build GRCh38
rs776495757 Chr.2: 112657055-112657055 on Build GRCh38
rs773846621 Chr.2: 112656959-112656959 on Build GRCh38
rs377365741 Chr.2: 112657084-112657084 on Build GRCh38
rs778560261 Chr.2: 112657020-112657020 on Build GRCh38
rs746582790 Chr.2: 112657137-112657137 on Build GRCh38
rs755243713 Chr.2: 112657074-112657074 on Build GRCh38
rs779616483 Chr.2: 112657113-112657113 on Build GRCh38
rs531668777 Chr.2: 112657103-112657103 on Build GRCh38
rs781623205 Chr.2: 112656988-112656988 on Build GRCh38

View Full Product Details