Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143818454-143818689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PUF60
Entrez Gene ID
22827
Gene Name
poly(U) binding splicing factor 60
Gene Aliases
FIR, RoBPI, SIAHBP1, VRJS
UniGene
Hs.521924
Gene Chromosome Location
Chr.8: 143816344-143829386 on Build GRCh38
Transcripts
NM_001136033.2 NM_001271096.1 NM_001271097.1 NM_001271098.1 NM_001271099.1 NM_001271100.1 NM_014281.4 NM_078480.2 XM_011516929.2 XM_011516930.2 XM_017013234.1 XM_017013235.1 XM_017013236.1 XM_017013237.1 XM_017013238.1 XM_017013239.1 XM_017013240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3897536 c.419T>C p.L140P Chr.8: 143818464-143818464 on Build GRCh38
6760309 c.391G>A p.V131I Chr.8: 143818492-143818492 on Build GRCh38
4150598 c.390C>T p.R130R Chr.8: 143818493-143818493 on Build GRCh38
5408982 c.367C>T p.R123W Chr.8: 143818516-143818516 on Build GRCh38
6516744 c.399G>T p.V133V Chr.8: 143818484-143818484 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008964688 Chr.8: 143818634-143818634 on Build GRCh38
rs773430854 Chr.8: 143818552-143818552 on Build GRCh38
rs780699513 Chr.8: 143818487-143818487 on Build GRCh38
rs748433680 Chr.8: 143818540-143818540 on Build GRCh38
rs955299182 Chr.8: 143818631-143818631 on Build GRCh38
rs780390938 Chr.8: 143818511-143818511 on Build GRCh38
rs987424466 Chr.8: 143818633-143818633 on Build GRCh38
rs542357649 Chr.8: 143818643-143818643 on Build GRCh38
rs995522325 Chr.8: 143818660-143818660 on Build GRCh38
rs977913183 Chr.8: 143818629-143818629 on Build GRCh38
rs377214624 Chr.8: 143818579-143818579 on Build GRCh38
rs1026546442 Chr.8: 143818510-143818510 on Build GRCh38
rs756686350 Chr.8: 143818481-143818481 on Build GRCh38
rs7833632 Chr.8: 143818586-143818586 on Build GRCh38
rs745797494 Chr.8: 143818553-143818553 on Build GRCh38
rs890549627 Chr.8: 143818620-143818620 on Build GRCh38
rs922482911 Chr.8: 143818630-143818630 on Build GRCh38
rs761535408 Chr.8: 143818578-143818578 on Build GRCh38
rs748157247 Chr.8: 143818639-143818639 on Build GRCh38
rs762700045 Chr.8: 143818568-143818568 on Build GRCh38
rs562217993 Chr.8: 143818570-143818570 on Build GRCh38
rs910581509 Chr.8: 143818656-143818656 on Build GRCh38
rs773830015 Chr.8: 143818575-143818575 on Build GRCh38
rs750868114 Chr.8: 143818493-143818493 on Build GRCh38
rs772221323 Chr.8: 143818544-143818544 on Build GRCh38
rs749670489 Chr.8: 143818512-143818512 on Build GRCh38
rs372778526 Chr.8: 143818569-143818569 on Build GRCh38
rs369414220 Chr.8: 143818530-143818530 on Build GRCh38
rs879094365 Chr.8: 143818665-143818665 on Build GRCh38
rs966917352 Chr.8: 143818627-143818627 on Build GRCh38
rs775405689 Chr.8: 143818562-143818562 on Build GRCh38
rs749999625 Chr.8: 143818584-143818584 on Build GRCh38
rs768900134 Chr.8: 143818529-143818529 on Build GRCh38
rs771441852 Chr.8: 143818547-143818548 on Build GRCh38
rs573398282 Chr.8: 143818561-143818561 on Build GRCh38

View Full Product Details