Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21703323-21703596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL1
Entrez Gene ID
29799
Gene Name
yippee like 1
Gene Aliases
FKSG3
UniGene
Hs.517436
Gene Chromosome Location
Chr.22: 21697536-21735834 on Build GRCh38
Transcripts
NM_013313.4 NR_130910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5366571 c.219G>A p.A73A Chr.22: 21703421-21703421 on Build GRCh38
5393214 c.238G>A p.E80K Chr.22: 21703402-21703402 on Build GRCh38
7145328 c.190G>C p.E64Q Chr.22: 21703450-21703450 on Build GRCh38
5057705 c.178G>A p.G60S Chr.22: 21703462-21703462 on Build GRCh38
4102900 c.225C>T p.A75A Chr.22: 21703415-21703415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760105105 Chr.22: 21703519-21703520 on Build GRCh38
rs766652165 Chr.22: 21703463-21703463 on Build GRCh38
rs772409705 Chr.22: 21703428-21703428 on Build GRCh38
rs781218002 Chr.22: 21703484-21703484 on Build GRCh38
rs367906237 Chr.22: 21703347-21703347 on Build GRCh38
rs375936363 Chr.22: 21703512-21703512 on Build GRCh38
rs773575429 Chr.22: 21703433-21703433 on Build GRCh38
rs777632162 Chr.22: 21703344-21703344 on Build GRCh38
rs759586286 Chr.22: 21703468-21703468 on Build GRCh38
rs776881975 Chr.22: 21703519-21703519 on Build GRCh38
rs750248361 Chr.22: 21703491-21703491 on Build GRCh38
rs747549650 Chr.22: 21703511-21703511 on Build GRCh38
rs868034833 Chr.22: 21703355-21703355 on Build GRCh38
rs563850610 Chr.22: 21703515-21703516 on Build GRCh38
rs918212256 Chr.22: 21703487-21703487 on Build GRCh38
rs768188883 Chr.22: 21703403-21703403 on Build GRCh38
rs765850505 Chr.22: 21703514-21703515 on Build GRCh38
rs942455013 Chr.22: 21703564-21703564 on Build GRCh38
rs771265338 Chr.22: 21703518-21703518 on Build GRCh38
rs752683534 Chr.22: 21703482-21703482 on Build GRCh38
rs765332759 Chr.22: 21703472-21703472 on Build GRCh38
rs140503337 Chr.22: 21703400-21703400 on Build GRCh38
rs367940049 Chr.22: 21703514-21703514 on Build GRCh38
rs762413372 Chr.22: 21703421-21703421 on Build GRCh38
rs774758879 Chr.22: 21703415-21703415 on Build GRCh38
rs757245735 Chr.22: 21703483-21703483 on Build GRCh38
rs56658400 Chr.22: 21703523-21703524 on Build GRCh38
rs1002177027 Chr.22: 21703567-21703567 on Build GRCh38
rs990942660 Chr.22: 21703389-21703389 on Build GRCh38
rs372739031 Chr.22: 21703503-21703503 on Build GRCh38
rs779788033 Chr.22: 21703386-21703386 on Build GRCh38
rs760277142 Chr.22: 21703508-21703509 on Build GRCh38
rs1025444549 Chr.22: 21703537-21703537 on Build GRCh38
rs772872249 Chr.22: 21703507-21703527 on Build GRCh38
rs368486684 Chr.22: 21703502-21703502 on Build GRCh38
rs746945826 Chr.22: 21703346-21703346 on Build GRCh38
rs781168732 Chr.22: 21703356-21703356 on Build GRCh38
rs181575531 Chr.22: 21703513-21703513 on Build GRCh38
rs375164386 Chr.22: 21703495-21703495 on Build GRCh38
rs745609512 Chr.22: 21703370-21703370 on Build GRCh38
rs577315906 Chr.22: 21703498-21703498 on Build GRCh38
rs754044706 Chr.22: 21703466-21703466 on Build GRCh38
rs945146158 Chr.22: 21703429-21703429 on Build GRCh38
rs867363252 Chr.22: 21703522-21703522 on Build GRCh38
rs371636873 Chr.22: 21703382-21703382 on Build GRCh38

View Full Product Details