Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 5400853-5401118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBAL3
Entrez Gene ID
79861
Gene Name
tubulin alpha like 3
Gene Aliases
-
UniGene
Hs.163079
Gene Chromosome Location
Chr.10: 5393098-5404830 on Build GRCh38
Transcripts
NM_001171864.1 NM_024803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4766310 c.23A>G p.H8R Chr.10: 5401068-5401068 on Build GRCh38
6224555 c.168C>T p.F56F Chr.10: 5400923-5400923 on Build GRCh38
1702008 c.133G>A p.E45K Chr.10: 5400958-5400958 on Build GRCh38
6221279 c.44A>G p.Q15R Chr.10: 5401047-5401047 on Build GRCh38
6217470 c.102C>T p.G34G Chr.10: 5400989-5400989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782478971 Chr.10: 5401049-5401049 on Build GRCh38
rs782092117 Chr.10: 5400986-5400986 on Build GRCh38
rs782639744 Chr.10: 5400922-5400922 on Build GRCh38
rs150701455 Chr.10: 5401063-5401063 on Build GRCh38
rs958208277 Chr.10: 5400970-5400970 on Build GRCh38
rs782561379 Chr.10: 5400912-5400912 on Build GRCh38
rs782467469 Chr.10: 5401039-5401039 on Build GRCh38
rs201141984 Chr.10: 5401017-5401017 on Build GRCh38
rs782687182 Chr.10: 5401036-5401036 on Build GRCh38
rs147611249 Chr.10: 5400988-5400988 on Build GRCh38
rs781954116 Chr.10: 5401069-5401069 on Build GRCh38
rs1013676766 Chr.10: 5400886-5400886 on Build GRCh38
rs782493050 Chr.10: 5401035-5401035 on Build GRCh38
rs782199994 Chr.10: 5400952-5400952 on Build GRCh38
rs574462898 Chr.10: 5401009-5401009 on Build GRCh38
rs782134392 Chr.10: 5401079-5401079 on Build GRCh38
rs781860870 Chr.10: 5400923-5400924 on Build GRCh38
rs370481213 Chr.10: 5400958-5400958 on Build GRCh38
rs781882162 Chr.10: 5400903-5400903 on Build GRCh38
rs782092896 Chr.10: 5401024-5401024 on Build GRCh38
rs782311341 Chr.10: 5400955-5400955 on Build GRCh38
rs782816633 Chr.10: 5400890-5400890 on Build GRCh38
rs782671416 Chr.10: 5400936-5400936 on Build GRCh38
rs142953109 Chr.10: 5400913-5400913 on Build GRCh38
rs376134914 Chr.10: 5401084-5401084 on Build GRCh38
rs782479600 Chr.10: 5400918-5400918 on Build GRCh38
rs376910379 Chr.10: 5400989-5400989 on Build GRCh38
rs139186059 Chr.10: 5400884-5400884 on Build GRCh38
rs140117932 Chr.10: 5401066-5401066 on Build GRCh38
rs182163936 Chr.10: 5400888-5400888 on Build GRCh38
rs782249050 Chr.10: 5401060-5401060 on Build GRCh38
rs782059123 Chr.10: 5400874-5400874 on Build GRCh38
rs782031437 Chr.10: 5400902-5400902 on Build GRCh38
rs781869949 Chr.10: 5401027-5401027 on Build GRCh38
rs782147481 Chr.10: 5401088-5401088 on Build GRCh38
rs782449341 Chr.10: 5400904-5400904 on Build GRCh38
rs148158899 Chr.10: 5401037-5401037 on Build GRCh38
rs782743908 Chr.10: 5400877-5400877 on Build GRCh38
rs199802708 Chr.10: 5400963-5400963 on Build GRCh38
rs187231850 Chr.10: 5401065-5401065 on Build GRCh38
rs34911632 Chr.10: 5400945-5400946 on Build GRCh38
rs534913397 Chr.10: 5400923-5400923 on Build GRCh38
rs556747045 Chr.10: 5401064-5401064 on Build GRCh38
rs782729778 Chr.10: 5401026-5401026 on Build GRCh38
rs113521419 Chr.10: 5400942-5400942 on Build GRCh38
rs377249917 Chr.10: 5400929-5400929 on Build GRCh38
rs782141044 Chr.10: 5400996-5400996 on Build GRCh38

View Full Product Details