Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113393885-113394158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDS
Entrez Gene ID
10993
Gene Name
serine dehydratase
Gene Aliases
SDH
UniGene
Hs.439023
Gene Chromosome Location
Chr.12: 113392445-113403887 on Build GRCh38
Transcripts
NM_006843.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6773932 c.670G>T p.G224C Chr.12: 113394000-113394000 on Build GRCh38
935434 c.735T>C p.V245V Chr.12: 113393935-113393935 on Build GRCh38
6773937 c.726C>A p.F242L Chr.12: 113393944-113393944 on Build GRCh38
7389069 c.715C>T p.H239Y Chr.12: 113393955-113393955 on Build GRCh38
4396125 c.693G>A p.Q231Q Chr.12: 113393977-113393977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780619588 Chr.12: 113393998-113393998 on Build GRCh38
rs914127591 Chr.12: 113394127-113394127 on Build GRCh38
rs79165243 Chr.12: 113394076-113394076 on Build GRCh38
rs749353246 Chr.12: 113394063-113394063 on Build GRCh38
rs370861483 Chr.12: 113394035-113394035 on Build GRCh38
rs764891233 Chr.12: 113394004-113394004 on Build GRCh38
rs1020035176 Chr.12: 113393967-113393967 on Build GRCh38
rs975094824 Chr.12: 113394054-113394054 on Build GRCh38
rs758548948 Chr.12: 113393955-113393955 on Build GRCh38
rs201592901 Chr.12: 113394066-113394066 on Build GRCh38
rs940833693 Chr.12: 113394104-113394104 on Build GRCh38
rs764085895 Chr.12: 113393989-113393989 on Build GRCh38
rs776963482 Chr.12: 113394132-113394132 on Build GRCh38
rs199917625 Chr.12: 113393916-113393916 on Build GRCh38
rs978598372 Chr.12: 113394003-113394003 on Build GRCh38
rs1050062 Chr.12: 113393982-113393982 on Build GRCh38
rs760583748 Chr.12: 113393987-113393987 on Build GRCh38
rs763558202 Chr.12: 113393910-113393910 on Build GRCh38
rs557767195 Chr.12: 113394065-113394065 on Build GRCh38
rs376262267 Chr.12: 113394047-113394047 on Build GRCh38
rs560043234 Chr.12: 113394080-113394080 on Build GRCh38
rs758015294 Chr.12: 113394019-113394019 on Build GRCh38
rs774669339 Chr.12: 113393969-113393969 on Build GRCh38
rs771073696 Chr.12: 113393953-113393953 on Build GRCh38
rs866232822 Chr.12: 113393972-113393972 on Build GRCh38
rs749325165 Chr.12: 113393951-113393951 on Build GRCh38
rs767928134 Chr.12: 113394026-113394026 on Build GRCh38
rs899233471 Chr.12: 113394122-113394122 on Build GRCh38
rs111912553 Chr.12: 113393908-113393908 on Build GRCh38
rs778287697 Chr.12: 113394052-113394052 on Build GRCh38
rs1040654106 Chr.12: 113394125-113394125 on Build GRCh38
rs753372802 Chr.12: 113393911-113393911 on Build GRCh38
rs777783554 Chr.12: 113393937-113393937 on Build GRCh38
rs761326834 Chr.12: 113394000-113394000 on Build GRCh38
rs751933861 Chr.12: 113393925-113393925 on Build GRCh38
rs368480339 Chr.12: 113393983-113393983 on Build GRCh38
rs756584712 Chr.12: 113394041-113394041 on Build GRCh38
rs986146852 Chr.12: 113394088-113394090 on Build GRCh38
rs750036739 Chr.12: 113394010-113394010 on Build GRCh38
rs745693112 Chr.12: 113393974-113393974 on Build GRCh38
rs756631348 Chr.12: 113393930-113393930 on Build GRCh38
rs374014118 Chr.12: 113394044-113394044 on Build GRCh38
rs769131069 Chr.12: 113394130-113394130 on Build GRCh38
rs142172311 Chr.12: 113394105-113394105 on Build GRCh38
rs376660804 Chr.12: 113393929-113393929 on Build GRCh38
rs371979944 Chr.12: 113393997-113393997 on Build GRCh38
rs908010544 Chr.12: 113394088-113394088 on Build GRCh38

View Full Product Details