Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 52132854-52133123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST18
Entrez Gene ID
9705
Gene Name
ST18, C2H2C-type zinc finger
Gene Aliases
NZF3, ZC2H2C3, ZC2HC10, ZNF387
UniGene
Hs.655499
Gene Chromosome Location
Chr.8: 52110832-52409879 on Build GRCh38
Transcripts
NM_014682.2 XM_006716487.1 XM_011517629.1 XM_011517631.1 XM_011517632.1 XM_011517633.1 XM_011517634.1 XM_011517635.1 XM_011517636.2 XM_011517637.1 XM_011517638.2 XM_011517641.1 XM_011517642.1 XM_017014047.1 XM_017014048.1 XM_017014049.1 XM_017014050.1 XM_017014051.1 XM_017014052.1 XM_017014053.1 XM_017014054.1 XM_017014055.1 XM_017014056.1 XM_017014057.1 XM_017014058.1 XM_017014059.1 XM_017014060.1 XM_017014061.1 XM_017014062.1 XM_017014063.1 XM_017014064.1 XM_017014065.1 XM_017014066.1 XM_017014067.1 XM_017014068.1 XM_017014069.1 XM_017014070.1 XM_017014071.1 XM_017014072.1 XM_017014073.1 XM_017014074.1 XM_017014075.1 XM_017014076.1 XM_017014077.1 XM_017014078.1 XM_017014079.1 XM_017014080.1 XM_017014081.1 XM_017014082.1 XM_017014083.1 XM_017014084.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227664 c.2431G>T p.D811Y Chr.8: 52133070-52133070 on Build GRCh38
6181322 c.2437G>T p.E813* Chr.8: 52133064-52133064 on Build GRCh38
5063257 c.2380C>T p.R794C Chr.8: 52133121-52133121 on Build GRCh38
5463728 c.2394T>C p.G798G Chr.8: 52133107-52133107 on Build GRCh38
2718647 c.2381G>A p.R794H Chr.8: 52133120-52133120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758484650 Chr.8: 52133033-52133033 on Build GRCh38
rs1020274352 Chr.8: 52133022-52133022 on Build GRCh38
rs866522636 Chr.8: 52133082-52133082 on Build GRCh38
rs1037250118 Chr.8: 52132971-52132971 on Build GRCh38
rs755201614 Chr.8: 52133060-52133060 on Build GRCh38
rs377438682 Chr.8: 52132987-52132987 on Build GRCh38
rs1057411936 Chr.8: 52132893-52132893 on Build GRCh38
rs541058024 Chr.8: 52132938-52132938 on Build GRCh38
rs372147635 Chr.8: 52133049-52133049 on Build GRCh38
rs760273210 Chr.8: 52133059-52133059 on Build GRCh38
rs1012715822 Chr.8: 52132940-52132940 on Build GRCh38
rs965591896 Chr.8: 52133025-52133025 on Build GRCh38
rs752477054 Chr.8: 52133016-52133016 on Build GRCh38
rs367908168 Chr.8: 52133017-52133017 on Build GRCh38
rs1007329646 Chr.8: 52133068-52133068 on Build GRCh38
rs552686393 Chr.8: 52132969-52132969 on Build GRCh38
rs975599641 Chr.8: 52133085-52133085 on Build GRCh38
rs764184960 Chr.8: 52133020-52133020 on Build GRCh38
rs897453370 Chr.8: 52132981-52132981 on Build GRCh38
rs965869967 Chr.8: 52133091-52133091 on Build GRCh38
rs868627366 Chr.8: 52133077-52133077 on Build GRCh38
rs769396714 Chr.8: 52133029-52133030 on Build GRCh38
rs887421796 Chr.8: 52132999-52132999 on Build GRCh38
rs770269660 Chr.8: 52133096-52133096 on Build GRCh38
rs895681006 Chr.8: 52132923-52132923 on Build GRCh38
rs748577075 Chr.8: 52133093-52133093 on Build GRCh38
rs767325666 Chr.8: 52133015-52133015 on Build GRCh38
rs747203743 Chr.8: 52133052-52133052 on Build GRCh38
rs35633588 Chr.8: 52133007-52133007 on Build GRCh38
rs374833831 Chr.8: 52133089-52133089 on Build GRCh38
rs759136089 Chr.8: 52133006-52133006 on Build GRCh38
rs996155449 Chr.8: 52132985-52132985 on Build GRCh38
rs201099995 Chr.8: 52133028-52133028 on Build GRCh38
rs905288178 Chr.8: 52132951-52132951 on Build GRCh38
rs951457698 Chr.8: 52132995-52132995 on Build GRCh38
rs1022729198 Chr.8: 52132947-52132947 on Build GRCh38

View Full Product Details