Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 122803341-122803592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX2
Entrez Gene ID
6643
Gene Name
sorting nexin 2
Gene Aliases
TRG-9
UniGene
Hs.125352
Gene Chromosome Location
Chr.5: 122774996-122834539 on Build GRCh38
Transcripts
NM_001278199.1 NM_003100.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7326771 c.502-6_502-5ins18 p.? Chr.5: 122803466-122803467 on Build GRCh38
3135111 c.556G>A p.D186N Chr.5: 122803526-122803526 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545609024 Chr.5: 122803377-122803377 on Build GRCh38
rs147977984 Chr.5: 122803525-122803525 on Build GRCh38
rs1018972486 Chr.5: 122803560-122803560 on Build GRCh38
rs771488440 Chr.5: 122803492-122803492 on Build GRCh38
rs751566451 Chr.5: 122803485-122803485 on Build GRCh38
rs767892519 Chr.5: 122803516-122803516 on Build GRCh38
rs904118020 Chr.5: 122803566-122803566 on Build GRCh38
rs772917529 Chr.5: 122803455-122803455 on Build GRCh38
rs1050822235 Chr.5: 122803413-122803413 on Build GRCh38
rs759124364 Chr.5: 122803481-122803481 on Build GRCh38
rs764928550 Chr.5: 122803482-122803482 on Build GRCh38
rs746939515 Chr.5: 122803463-122803463 on Build GRCh38
rs563922246 Chr.5: 122803388-122803388 on Build GRCh38
rs761196086 Chr.5: 122803526-122803526 on Build GRCh38
rs111277526 Chr.5: 122803522-122803522 on Build GRCh38
rs748426177 Chr.5: 122803437-122803437 on Build GRCh38
rs753855706 Chr.5: 122803538-122803538 on Build GRCh38
rs766827774 Chr.5: 122803528-122803528 on Build GRCh38
rs755035800 Chr.5: 122803545-122803545 on Build GRCh38
rs573073599 Chr.5: 122803382-122803382 on Build GRCh38
rs755123899 Chr.5: 122803425-122803425 on Build GRCh38
rs367719650 Chr.5: 122803428-122803428 on Build GRCh38
rs762764284 Chr.5: 122803501-122803501 on Build GRCh38
rs776238366 Chr.5: 122803471-122803471 on Build GRCh38
rs770793417 Chr.5: 122803465-122803465 on Build GRCh38
rs147769824 Chr.5: 122803399-122803399 on Build GRCh38
rs771895859 Chr.5: 122803454-122803454 on Build GRCh38

View Full Product Details