Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58471983-58472214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF324
Entrez Gene ID
25799
Gene Name
zinc finger protein 324
Gene Aliases
ZF5128, ZNF324A
UniGene
Hs.515660
Gene Chromosome Location
Chr.19: 58467045-58473578 on Build GRCh38
Transcripts
NM_014347.2 XM_005258713.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713041 c.1541C>T p.A514V Chr.19: 58472033-58472033 on Build GRCh38
7149409 c.1500G>C p.Q500H Chr.19: 58471992-58471992 on Build GRCh38
4634481 c.1571C>T p.A524V Chr.19: 58472063-58472063 on Build GRCh38
6829335 c.1553C>A p.P518H Chr.19: 58472045-58472045 on Build GRCh38
5057038 c.1592C>T p.A531V Chr.19: 58472084-58472084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530266629 Chr.19: 58472131-58472131 on Build GRCh38
rs751647991 Chr.19: 58472108-58472108 on Build GRCh38
rs763252728 Chr.19: 58472178-58472178 on Build GRCh38
rs773578671 Chr.19: 58472086-58472086 on Build GRCh38
rs939182679 Chr.19: 58472047-58472047 on Build GRCh38
rs762648210 Chr.19: 58472020-58472020 on Build GRCh38
rs761119095 Chr.19: 58472041-58472041 on Build GRCh38
rs759787884 Chr.19: 58472128-58472128 on Build GRCh38
rs754229505 Chr.19: 58472075-58472077 on Build GRCh38
rs756008886 Chr.19: 58472110-58472110 on Build GRCh38
rs138554661 Chr.19: 58472038-58472038 on Build GRCh38
rs370864217 Chr.19: 58472163-58472163 on Build GRCh38
rs142263975 Chr.19: 58472126-58472126 on Build GRCh38
rs113119239 Chr.19: 58472074-58472074 on Build GRCh38
rs372264496 Chr.19: 58472015-58472015 on Build GRCh38
rs148766951 Chr.19: 58472100-58472100 on Build GRCh38
rs771637415 Chr.19: 58472120-58472120 on Build GRCh38
rs184685134 Chr.19: 58472176-58472176 on Build GRCh38
rs757381183 Chr.19: 58472151-58472151 on Build GRCh38
rs761200510 Chr.19: 58472085-58472085 on Build GRCh38
rs1005962679 Chr.19: 58472145-58472145 on Build GRCh38
rs11673703 Chr.19: 58472182-58472182 on Build GRCh38
rs758028947 Chr.19: 58472162-58472162 on Build GRCh38
rs759988285 Chr.19: 58472157-58472158 on Build GRCh38
rs192677679 Chr.19: 58472165-58472165 on Build GRCh38
rs749063610 Chr.19: 58472113-58472113 on Build GRCh38
rs751594468 Chr.19: 58472140-58472140 on Build GRCh38
rs763442788 Chr.19: 58472130-58472130 on Build GRCh38
rs55875034 Chr.19: 58472191-58472191 on Build GRCh38
rs768306218 Chr.19: 58472024-58472024 on Build GRCh38
rs149270714 Chr.19: 58472046-58472046 on Build GRCh38
rs367682553 Chr.19: 58472009-58472009 on Build GRCh38
rs146385895 Chr.19: 58472154-58472154 on Build GRCh38
rs545395456 Chr.19: 58472064-58472064 on Build GRCh38
rs761410628 Chr.19: 58472034-58472034 on Build GRCh38
rs765991531 Chr.19: 58472003-58472018 on Build GRCh38
rs964729780 Chr.19: 58472157-58472157 on Build GRCh38
rs913336836 Chr.19: 58472103-58472103 on Build GRCh38
rs764222713 Chr.19: 58472136-58472136 on Build GRCh38
rs754269217 Chr.19: 58472043-58472043 on Build GRCh38
rs3764527 Chr.19: 58472177-58472177 on Build GRCh38
rs377733639 Chr.19: 58472091-58472091 on Build GRCh38
rs540503836 Chr.19: 58472005-58472005 on Build GRCh38
rs768148627 Chr.19: 58472078-58472078 on Build GRCh38
rs755183892 Chr.19: 58472045-58472045 on Build GRCh38
rs945275650 Chr.19: 58472069-58472069 on Build GRCh38
rs201793822 Chr.19: 58472090-58472090 on Build GRCh38
rs1047573797 Chr.19: 58472134-58472134 on Build GRCh38
rs749115951 Chr.19: 58472077-58472077 on Build GRCh38
rs766275664 Chr.19: 58472152-58472152 on Build GRCh38
rs201171317 Chr.19: 58472016-58472016 on Build GRCh38
rs775115485 Chr.19: 58472017-58472017 on Build GRCh38
rs772781917 Chr.19: 58472124-58472124 on Build GRCh38
rs763173845 Chr.19: 58472135-58472135 on Build GRCh38
rs777962869 Chr.19: 58472106-58472106 on Build GRCh38
rs778712737 Chr.19: 58472159-58472159 on Build GRCh38
rs777324700 Chr.19: 58472127-58472127 on Build GRCh38
rs61731807 Chr.19: 58472081-58472081 on Build GRCh38
rs200150466 Chr.19: 58472042-58472042 on Build GRCh38
rs773698937 Chr.19: 58472033-58472033 on Build GRCh38
rs747900691 Chr.19: 58472160-58472160 on Build GRCh38
rs746442765 Chr.19: 58472171-58472171 on Build GRCh38
rs886204171 Chr.19: 58472143-58472143 on Build GRCh38
rs769559083 Chr.19: 58472067-58472067 on Build GRCh38
rs376032280 Chr.19: 58472184-58472184 on Build GRCh38
rs752669190 Chr.19: 58472053-58472053 on Build GRCh38
rs747893723 Chr.19: 58472118-58472118 on Build GRCh38
rs758837872 Chr.19: 58472057-58472057 on Build GRCh38
rs57153006 Chr.19: 58472148-58472148 on Build GRCh38
rs779763030 Chr.19: 58472072-58472072 on Build GRCh38
rs758401311 Chr.19: 58472101-58472101 on Build GRCh38
rs765768397 Chr.19: 58472177-58472179 on Build GRCh38
rs927267092 Chr.19: 58472111-58472111 on Build GRCh38
rs758553002 Chr.19: 58472061-58472061 on Build GRCh38
rs767891137 Chr.19: 58472037-58472037 on Build GRCh38
rs781210717 Chr.19: 58472008-58472008 on Build GRCh38
rs561509368 Chr.19: 58472112-58472112 on Build GRCh38
rs965017648 Chr.19: 58472056-58472056 on Build GRCh38
rs748264086 Chr.19: 58472004-58472004 on Build GRCh38
rs754718188 Chr.19: 58472155-58472155 on Build GRCh38
rs745869560 Chr.19: 58472065-58472065 on Build GRCh38
rs778428124 Chr.19: 58472117-58472117 on Build GRCh38
rs764282417 Chr.19: 58472179-58472179 on Build GRCh38
rs374200034 Chr.19: 58472167-58472167 on Build GRCh38

View Full Product Details