Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 7170855-7171128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFMBT2
Entrez Gene ID
57713
Gene Name
Scm-like with four mbt domains 2
Gene Aliases
-
UniGene
Hs.407983
Gene Chromosome Location
Chr.10: 7158624-7411486 on Build GRCh38
Transcripts
NM_001018039.1 NM_001029880.2 XM_005252551.2 XM_006717490.2 XM_011519611.1 XM_017016466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5749690 c.2477T>G p.V826G Chr.10: 7170995-7170995 on Build GRCh38
4015480 c.2528A>C p.K843T Chr.10: 7170944-7170944 on Build GRCh38
1165882 c.2462C>T p.P821L Chr.10: 7171010-7171010 on Build GRCh38
6358423 c.2544+6G>A p.? Chr.10: 7170922-7170922 on Build GRCh38
5626344 c.2485G>A p.V829M Chr.10: 7170987-7170987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138548214 Chr.10: 7170895-7170895 on Build GRCh38
rs754426857 Chr.10: 7171075-7171075 on Build GRCh38
rs373045151 Chr.10: 7170914-7170914 on Build GRCh38
rs905345550 Chr.10: 7170885-7170885 on Build GRCh38
rs745846779 Chr.10: 7171108-7171108 on Build GRCh38
rs779290101 Chr.10: 7170879-7170879 on Build GRCh38
rs890183721 Chr.10: 7171032-7171032 on Build GRCh38
rs771544652 Chr.10: 7171032-7171033 on Build GRCh38
rs760958777 Chr.10: 7171068-7171068 on Build GRCh38
rs201629053 Chr.10: 7170970-7170970 on Build GRCh38
rs777179242 Chr.10: 7171091-7171092 on Build GRCh38
rs373483982 Chr.10: 7171027-7171027 on Build GRCh38
rs761242244 Chr.10: 7170906-7170906 on Build GRCh38
rs746763707 Chr.10: 7171095-7171095 on Build GRCh38
rs748417020 Chr.10: 7170992-7170992 on Build GRCh38
rs201201333 Chr.10: 7171079-7171079 on Build GRCh38
rs758102258 Chr.10: 7170997-7170997 on Build GRCh38
rs1037657308 Chr.10: 7170984-7170984 on Build GRCh38
rs776275275 Chr.10: 7170907-7170907 on Build GRCh38
rs745336542 Chr.10: 7171041-7171041 on Build GRCh38
rs766859921 Chr.10: 7171070-7171070 on Build GRCh38
rs758710998 Chr.10: 7171089-7171089 on Build GRCh38
rs1038657235 Chr.10: 7171039-7171039 on Build GRCh38
rs897661040 Chr.10: 7170988-7170988 on Build GRCh38
rs756703291 Chr.10: 7170924-7170924 on Build GRCh38
rs769325050 Chr.10: 7171051-7171051 on Build GRCh38
rs200964369 Chr.10: 7170911-7170911 on Build GRCh38
rs766799863 Chr.10: 7170952-7170952 on Build GRCh38
rs118074539 Chr.10: 7171052-7171052 on Build GRCh38
rs753910392 Chr.10: 7170960-7170960 on Build GRCh38
rs752372137 Chr.10: 7170913-7170913 on Build GRCh38
rs369892171 Chr.10: 7170912-7170912 on Build GRCh38
rs147608900 Chr.10: 7171009-7171009 on Build GRCh38
rs929117063 Chr.10: 7170991-7170991 on Build GRCh38
rs747542096 Chr.10: 7171033-7171035 on Build GRCh38
rs773558275 Chr.10: 7171065-7171065 on Build GRCh38
rs368746547 Chr.10: 7171064-7171064 on Build GRCh38
rs771928501 Chr.10: 7170903-7170903 on Build GRCh38
rs757027886 Chr.10: 7171097-7171097 on Build GRCh38
rs192259966 Chr.10: 7171093-7171093 on Build GRCh38
rs750875082 Chr.10: 7170923-7170923 on Build GRCh38
rs142664169 Chr.10: 7171106-7171106 on Build GRCh38
rs529869842 Chr.10: 7170883-7170884 on Build GRCh38
rs142215982 Chr.10: 7171038-7171038 on Build GRCh38
rs376102425 Chr.10: 7171088-7171088 on Build GRCh38
rs1022746814 Chr.10: 7170881-7170881 on Build GRCh38
rs762644248 Chr.10: 7171055-7171055 on Build GRCh38
rs765453495 Chr.10: 7171086-7171086 on Build GRCh38
rs1014388059 Chr.10: 7171094-7171094 on Build GRCh38
rs767965917 Chr.10: 7170917-7170917 on Build GRCh38
rs767761584 Chr.10: 7171061-7171061 on Build GRCh38
rs779236687 Chr.10: 7170976-7170976 on Build GRCh38
rs761338317 Chr.10: 7171031-7171031 on Build GRCh38
rs1024904853 Chr.10: 7171103-7171103 on Build GRCh38

View Full Product Details