Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35709095-35709368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBM39
Entrez Gene ID
9584
Gene Name
RNA binding motif protein 39
Gene Aliases
CAPER, CAPERalpha, FSAP59, HCC1, RNPC2
UniGene
Hs.282901
Gene Chromosome Location
Chr.20: 35703609-35742336 on Build GRCh38
Transcripts
NM_001242599.1 NM_001242600.1 NM_001323422.1 NM_001323423.1 NM_001323424.1 NM_004902.3 NM_184234.2 NR_040722.1 NR_040723.1 NR_040724.1 XM_006723890.3 XM_006723891.3 XM_011529111.2 XM_017028138.1 XM_017028139.1 XM_017028140.1 XM_017028141.1 XM_017028142.1 XM_017028143.1 XM_017028144.1 XM_017028145.1 XM_017028146.1 XM_017028147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5037315 c.1225+2T>C p.? Chr.20: 35709222-35709222 on Build GRCh38
4097964 c.1217A>G p.Q406R Chr.20: 35709232-35709232 on Build GRCh38
221026 c.1190T>C p.I397T Chr.20: 35709259-35709259 on Build GRCh38
5833961 c.1184_1185delTT p.F395fs*13 Chr.20: 35709264-35709265 on Build GRCh38
5813182 c.1185T>G p.F395L Chr.20: 35709264-35709264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187695667 Chr.20: 35709184-35709184 on Build GRCh38
rs751530968 Chr.20: 35709261-35709261 on Build GRCh38
rs750215931 Chr.20: 35709208-35709208 on Build GRCh38
rs375806286 Chr.20: 35709207-35709207 on Build GRCh38
rs545518002 Chr.20: 35709211-35709211 on Build GRCh38
rs1055226632 Chr.20: 35709143-35709143 on Build GRCh38
rs781252997 Chr.20: 35709333-35709333 on Build GRCh38
rs755005003 Chr.20: 35709311-35709311 on Build GRCh38
rs904101672 Chr.20: 35709142-35709142 on Build GRCh38
rs896463659 Chr.20: 35709151-35709151 on Build GRCh38
rs562854635 Chr.20: 35709157-35709157 on Build GRCh38
rs753444976 Chr.20: 35709193-35709193 on Build GRCh38
rs112984367 Chr.20: 35709144-35709144 on Build GRCh38
rs781227754 Chr.20: 35709318-35709318 on Build GRCh38
rs1021183366 Chr.20: 35709162-35709162 on Build GRCh38
rs577948270 Chr.20: 35709205-35709205 on Build GRCh38
rs373895208 Chr.20: 35709240-35709240 on Build GRCh38
rs370414635 Chr.20: 35709217-35709217 on Build GRCh38
rs781748051 Chr.20: 35709215-35709216 on Build GRCh38
rs761492437 Chr.20: 35709204-35709204 on Build GRCh38
rs953242083 Chr.20: 35709329-35709329 on Build GRCh38

View Full Product Details