Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72359357-72359588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFAND5
Entrez Gene ID
7763
Gene Name
zinc finger AN1-type containing 5
Gene Aliases
ZA20D2, ZFAND5A, ZNF216
UniGene
Hs.406096
Gene Chromosome Location
Chr.9: 72351425-72365247 on Build GRCh38
Transcripts
NM_001102420.2 NM_001102421.2 NM_001278243.1 NM_001278244.1 NM_001278245.1 NM_006007.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4983521 c.295C>G p.Q99E Chr.9: 72359490-72359490 on Build GRCh38
6826321 c.270G>A p.V90V Chr.9: 72359515-72359515 on Build GRCh38
5455792 c.286C>T p.P96S Chr.9: 72359499-72359499 on Build GRCh38
6903165 c.342C>T p.T114T Chr.9: 72359443-72359443 on Build GRCh38
6400525 c.434C>T p.A145V Chr.9: 72359507-72359507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775426032 Chr.9: 72359440-72359440 on Build GRCh38
rs755295159 Chr.9: 72359508-72359508 on Build GRCh38
rs778350630 Chr.9: 72359541-72359541 on Build GRCh38
rs751936064 Chr.9: 72359505-72359505 on Build GRCh38
rs780474806 Chr.9: 72359504-72359504 on Build GRCh38
rs144844537 Chr.9: 72359446-72359446 on Build GRCh38
rs149441104 Chr.9: 72359431-72359431 on Build GRCh38
rs748941717 Chr.9: 72359517-72359517 on Build GRCh38
rs79349856 Chr.9: 72359500-72359500 on Build GRCh38
rs750503392 Chr.9: 72359490-72359490 on Build GRCh38
rs765285603 Chr.9: 72359475-72359475 on Build GRCh38
rs766366529 Chr.9: 72359450-72359450 on Build GRCh38
rs757474714 Chr.9: 72359455-72359455 on Build GRCh38
rs759589619 Chr.9: 72359557-72359558 on Build GRCh38
rs746547437 Chr.9: 72359401-72359401 on Build GRCh38
rs770603870 Chr.9: 72359518-72359518 on Build GRCh38
rs376158329 Chr.9: 72359441-72359441 on Build GRCh38
rs1002901496 Chr.9: 72359398-72359398 on Build GRCh38
rs1045755949 Chr.9: 72359524-72359524 on Build GRCh38
rs375153584 Chr.9: 72359403-72359403 on Build GRCh38
rs759449117 Chr.9: 72359406-72359406 on Build GRCh38
rs770386501 Chr.9: 72359402-72359402 on Build GRCh38
rs770847035 Chr.9: 72359407-72359408 on Build GRCh38
rs781587752 Chr.9: 72359510-72359510 on Build GRCh38
rs776441068 Chr.9: 72359524-72359525 on Build GRCh38
rs753969807 Chr.9: 72359449-72359449 on Build GRCh38
rs199782451 Chr.9: 72359506-72359506 on Build GRCh38
rs899362145 Chr.9: 72359435-72359435 on Build GRCh38
rs745306213 Chr.9: 72359556-72359556 on Build GRCh38

View Full Product Details