Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55718209-55718472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAGB
Entrez Gene ID
10325
Gene Name
Ras related GTP binding B
Gene Aliases
RAGB, bA465E19.1
UniGene
Hs.50282
Gene Chromosome Location
Chr.X: 55717677-55760209 on Build GRCh38
Transcripts
NM_006064.4 NM_016656.3 XM_005261973.4 XM_017029217.1 XM_017029218.1

SNP Detail

SNP ID Chromosome Location
rs368005933 Chr.X: 55718315-55718315 on Build GRCh38
rs912706975 Chr.X: 55718262-55718264 on Build GRCh38
rs1001312235 Chr.X: 55718346-55718346 on Build GRCh38
rs1024830248 Chr.X: 55718413-55718413 on Build GRCh38
rs763810949 Chr.X: 55718422-55718422 on Build GRCh38
rs970508617 Chr.X: 55718431-55718431 on Build GRCh38
rs764055473 Chr.X: 55718279-55718279 on Build GRCh38
rs780186698 Chr.X: 55718342-55718342 on Build GRCh38
rs372327982 Chr.X: 55718437-55718437 on Build GRCh38
rs769460101 Chr.X: 55718391-55718391 on Build GRCh38
rs750567996 Chr.X: 55718333-55718333 on Build GRCh38
rs909717094 Chr.X: 55718324-55718324 on Build GRCh38
rs749631631 Chr.X: 55718356-55718356 on Build GRCh38
rs186871522 Chr.X: 55718298-55718298 on Build GRCh38
rs892580164 Chr.X: 55718395-55718395 on Build GRCh38
rs757269584 Chr.X: 55718311-55718311 on Build GRCh38
rs756834227 Chr.X: 55718328-55718328 on Build GRCh38
rs143868192 Chr.X: 55718357-55718357 on Build GRCh38
rs369357317 Chr.X: 55718334-55718334 on Build GRCh38
rs770495932 Chr.X: 55718380-55718380 on Build GRCh38
rs1046428366 Chr.X: 55718400-55718400 on Build GRCh38
rs745423992 Chr.X: 55718384-55718384 on Build GRCh38
rs748674339 Chr.X: 55718363-55718363 on Build GRCh38
rs767590357 Chr.X: 55718321-55718321 on Build GRCh38
rs762761300 Chr.X: 55718405-55718405 on Build GRCh38
rs944296910 Chr.X: 55718314-55718314 on Build GRCh38
rs776271079 Chr.X: 55718383-55718383 on Build GRCh38
rs775040658 Chr.X: 55718397-55718397 on Build GRCh38

View Full Product Details