Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55424457-55424718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM165
Entrez Gene ID
55858
Gene Name
transmembrane protein 165
Gene Aliases
CDG2K, FT27, GDT1, TMPT27, TPARL
UniGene
Hs.479766
Gene Chromosome Location
Chr.4: 55395913-55453397 on Build GRCh38
Transcripts
NM_018475.4 NR_073070.1 XM_011534394.2 XM_017008412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753775 c.843G>C p.T281T Chr.4: 55424588-55424588 on Build GRCh38
1642492 c.873A>G p.I291M Chr.4: 55424618-55424618 on Build GRCh38
5666671 c.802G>T p.G268C Chr.4: 55424547-55424547 on Build GRCh38
294893 c.842C>T p.T281M Chr.4: 55424587-55424587 on Build GRCh38
1056163 c.846A>G p.G282G Chr.4: 55424591-55424591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765745912 Chr.4: 55424547-55424547 on Build GRCh38
rs372776107 Chr.4: 55424526-55424526 on Build GRCh38
rs531975275 Chr.4: 55424525-55424525 on Build GRCh38
rs770370528 Chr.4: 55424674-55424674 on Build GRCh38
rs866737576 Chr.4: 55424579-55424579 on Build GRCh38
rs112969727 Chr.4: 55424687-55424688 on Build GRCh38
rs202043214 Chr.4: 55424676-55424676 on Build GRCh38
rs751960423 Chr.4: 55424519-55424520 on Build GRCh38
rs904976758 Chr.4: 55424530-55424530 on Build GRCh38
rs764212150 Chr.4: 55424556-55424556 on Build GRCh38
rs781280352 Chr.4: 55424588-55424588 on Build GRCh38
rs1045244308 Chr.4: 55424543-55424543 on Build GRCh38
rs951824269 Chr.4: 55424669-55424669 on Build GRCh38
rs778746482 Chr.4: 55424622-55424622 on Build GRCh38
rs758596312 Chr.4: 55424614-55424614 on Build GRCh38
rs771840454 Chr.4: 55424651-55424651 on Build GRCh38
rs1016899022 Chr.4: 55424620-55424620 on Build GRCh38
rs199743518 Chr.4: 55424545-55424545 on Build GRCh38
rs369436814 Chr.4: 55424646-55424646 on Build GRCh38
rs772746415 Chr.4: 55424653-55424653 on Build GRCh38
rs1003602085 Chr.4: 55424540-55424540 on Build GRCh38
rs372064871 Chr.4: 55424561-55424561 on Build GRCh38
rs111449619 Chr.4: 55424644-55424644 on Build GRCh38
rs971950875 Chr.4: 55424609-55424609 on Build GRCh38
rs377397558 Chr.4: 55424685-55424685 on Build GRCh38
rs749268935 Chr.4: 55424592-55424592 on Build GRCh38
rs772835384 Chr.4: 55424492-55424492 on Build GRCh38
rs754879158 Chr.4: 55424598-55424598 on Build GRCh38
rs757310398 Chr.4: 55424576-55424576 on Build GRCh38
rs746634688 Chr.4: 55424659-55424659 on Build GRCh38
rs760164316 Chr.4: 55424686-55424686 on Build GRCh38
rs753083852 Chr.4: 55424555-55424555 on Build GRCh38

View Full Product Details