Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6753415-6753680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2D3A
Entrez Gene ID
10045
Gene Name
SH2 domain containing 3A
Gene Aliases
NSP1
UniGene
Hs.439645
Gene Chromosome Location
Chr.19: 6752160-6767513 on Build GRCh38
Transcripts
NM_005490.2 XM_005259467.1 XM_005259472.2 XM_006722602.1 XM_011527608.2 XM_011527609.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7285309 c.1460C>T p.A487V Chr.19: 6753566-6753566 on Build GRCh38
6506024 c.1568G>A p.R523Q Chr.19: 6753458-6753458 on Build GRCh38
6776973 c.1445A>G p.E482G Chr.19: 6753581-6753581 on Build GRCh38
4968038 c.1524C>T p.V508V Chr.19: 6753502-6753502 on Build GRCh38
6776969 c.1540T>C p.F514L Chr.19: 6753486-6753486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746129589 Chr.19: 6753653-6753653 on Build GRCh38
rs983799550 Chr.19: 6753505-6753505 on Build GRCh38
rs774494516 Chr.19: 6753455-6753455 on Build GRCh38
rs373836898 Chr.19: 6753578-6753578 on Build GRCh38
rs376684528 Chr.19: 6753517-6753517 on Build GRCh38
rs924748552 Chr.19: 6753536-6753536 on Build GRCh38
rs1051119137 Chr.19: 6753607-6753607 on Build GRCh38
rs756612734 Chr.19: 6753530-6753530 on Build GRCh38
rs754635882 Chr.19: 6753603-6753603 on Build GRCh38
rs376833079 Chr.19: 6753450-6753450 on Build GRCh38
rs746102756 Chr.19: 6753555-6753555 on Build GRCh38
rs373393251 Chr.19: 6753470-6753470 on Build GRCh38
rs751156704 Chr.19: 6753509-6753509 on Build GRCh38
rs767569299 Chr.19: 6753455-6753455 on Build GRCh38
rs1022074539 Chr.19: 6753491-6753491 on Build GRCh38
rs891142092 Chr.19: 6753636-6753636 on Build GRCh38
rs752279390 Chr.19: 6753606-6753606 on Build GRCh38
rs373443344 Chr.19: 6753486-6753486 on Build GRCh38
rs761800891 Chr.19: 6753465-6753465 on Build GRCh38
rs941904981 Chr.19: 6753526-6753526 on Build GRCh38
rs750188436 Chr.19: 6753499-6753499 on Build GRCh38
rs910291703 Chr.19: 6753585-6753585 on Build GRCh38
rs766156638 Chr.19: 6753582-6753582 on Build GRCh38
rs757766454 Chr.19: 6753646-6753646 on Build GRCh38
rs767458157 Chr.19: 6753593-6753593 on Build GRCh38
rs375639151 Chr.19: 6753510-6753510 on Build GRCh38
rs568976019 Chr.19: 6753651-6753651 on Build GRCh38
rs771305573 Chr.19: 6753444-6753444 on Build GRCh38
rs751325311 Chr.19: 6753500-6753500 on Build GRCh38
rs779705713 Chr.19: 6753506-6753506 on Build GRCh38
rs184596900 Chr.19: 6753522-6753522 on Build GRCh38
rs570757869 Chr.19: 6753539-6753539 on Build GRCh38
rs779410346 Chr.19: 6753652-6753652 on Build GRCh38
rs201959880 Chr.19: 6753594-6753594 on Build GRCh38
rs373630114 Chr.19: 6753448-6753448 on Build GRCh38
rs765190974 Chr.19: 6753576-6753576 on Build GRCh38
rs778435423 Chr.19: 6753634-6753634 on Build GRCh38
rs758150126 Chr.19: 6753502-6753502 on Build GRCh38
rs527327404 Chr.19: 6753520-6753520 on Build GRCh38
rs749743166 Chr.19: 6753637-6753637 on Build GRCh38
rs778380522 Chr.19: 6753534-6753534 on Build GRCh38
rs762752411 Chr.19: 6753581-6753581 on Build GRCh38
rs766974895 Chr.19: 6753604-6753604 on Build GRCh38
rs775621501 Chr.19: 6753565-6753565 on Build GRCh38
rs1035783655 Chr.19: 6753481-6753481 on Build GRCh38
rs568064266 Chr.19: 6753482-6753482 on Build GRCh38
rs370202096 Chr.19: 6753529-6753529 on Build GRCh38
rs765989877 Chr.19: 6753498-6753498 on Build GRCh38
rs771362495 Chr.19: 6753540-6753540 on Build GRCh38
rs772727234 Chr.19: 6753477-6753477 on Build GRCh38
rs988082584 Chr.19: 6753579-6753579 on Build GRCh38
rs778251511 Chr.19: 6753606-6753606 on Build GRCh38
rs369971495 Chr.19: 6753464-6753464 on Build GRCh38
rs774716402 Chr.19: 6753552-6753552 on Build GRCh38
rs148683690 Chr.19: 6753564-6753564 on Build GRCh38
rs555962781 Chr.19: 6753625-6753625 on Build GRCh38

View Full Product Details