Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100911046-100911279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC30A7
Entrez Gene ID
148867
Gene Name
solute carrier family 30 member 7
Gene Aliases
ZNT7, ZnT-7, ZnTL2
UniGene
Hs.533903
Gene Chromosome Location
Chr.1: 100896076-100996913 on Build GRCh38
Transcripts
NM_001144884.1 NM_133496.4 XM_011540779.2 XM_017000400.1 XM_017000401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4747673 c.357delT p.F121fs*11 Chr.1: 100911123-100911123 on Build GRCh38
6781478 c.332A>G p.N111S Chr.1: 100911098-100911098 on Build GRCh38
1953353 c.308C>T p.A103V Chr.1: 100911074-100911074 on Build GRCh38
1185528 c.356C>T p.A119V Chr.1: 100911122-100911122 on Build GRCh38
1472334 c.382G>C p.E128Q Chr.1: 100911148-100911148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs921105620 Chr.1: 100911143-100911143 on Build GRCh38
rs78872009 Chr.1: 100911094-100911094 on Build GRCh38
rs754303164 Chr.1: 100911074-100911074 on Build GRCh38
rs573856567 Chr.1: 100911075-100911075 on Build GRCh38
rs1036789697 Chr.1: 100911218-100911218 on Build GRCh38
rs745766132 Chr.1: 100911157-100911157 on Build GRCh38
rs765559215 Chr.1: 100911090-100911091 on Build GRCh38
rs975216786 Chr.1: 100911128-100911128 on Build GRCh38
rs779371671 Chr.1: 100911087-100911087 on Build GRCh38
rs773753433 Chr.1: 100911148-100911150 on Build GRCh38
rs542503042 Chr.1: 100911098-100911098 on Build GRCh38
rs763433869 Chr.1: 100911163-100911165 on Build GRCh38
rs986995054 Chr.1: 100911206-100911209 on Build GRCh38
rs770546458 Chr.1: 100911148-100911148 on Build GRCh38
rs115164886 Chr.1: 100911191-100911191 on Build GRCh38
rs780576042 Chr.1: 100911156-100911156 on Build GRCh38
rs371455415 Chr.1: 100911176-100911176 on Build GRCh38
rs909797780 Chr.1: 100911154-100911154 on Build GRCh38
rs777534332 Chr.1: 100911104-100911104 on Build GRCh38
rs767079367 Chr.1: 100911203-100911206 on Build GRCh38
rs753145079 Chr.1: 100911097-100911097 on Build GRCh38
rs746490759 Chr.1: 100911113-100911113 on Build GRCh38
rs151189633 Chr.1: 100911246-100911246 on Build GRCh38
rs751734792 Chr.1: 100911160-100911161 on Build GRCh38

View Full Product Details