Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87979931-87980204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPP1
Entrez Gene ID
6696
Gene Name
secreted phosphoprotein 1
Gene Aliases
BNSP, BSPI, ETA-1, OPN
UniGene
Hs.313
Gene Chromosome Location
Chr.4: 87975650-87983411 on Build GRCh38
Transcripts
NM_000582.2 NM_001040058.1 NM_001040060.1 NM_001251829.1 NM_001251830.1 XM_017008564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6862241 c.174+9A>C p.? Chr.4: 87980135-87980135 on Build GRCh38
1227421 c.172C>T p.Q58* Chr.4: 87980124-87980124 on Build GRCh38
5014439 c.152A>C p.K51T Chr.4: 87980104-87980104 on Build GRCh38
6836373 c.156G>T p.Q52H Chr.4: 87980108-87980108 on Build GRCh38
6789229 c.94-10delT p.? Chr.4: 87980036-87980036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902914665 Chr.4: 87979978-87979978 on Build GRCh38
rs768250066 Chr.4: 87979997-87979997 on Build GRCh38
rs2859697 Chr.4: 87980167-87980167 on Build GRCh38
rs1040822029 Chr.4: 87979953-87979953 on Build GRCh38
rs754853525 Chr.4: 87980096-87980096 on Build GRCh38
rs371560904 Chr.4: 87980032-87980032 on Build GRCh38
rs11544546 Chr.4: 87980092-87980092 on Build GRCh38
rs373731606 Chr.4: 87980079-87980079 on Build GRCh38
rs772876619 Chr.4: 87980143-87980143 on Build GRCh38
rs753342177 Chr.4: 87979957-87979957 on Build GRCh38
rs559127266 Chr.4: 87980179-87980179 on Build GRCh38
rs768909108 Chr.4: 87980035-87980036 on Build GRCh38
rs746605215 Chr.4: 87980169-87980169 on Build GRCh38
rs771141536 Chr.4: 87980162-87980162 on Build GRCh38
rs755968112 Chr.4: 87980072-87980072 on Build GRCh38
rs757338944 Chr.4: 87980061-87980061 on Build GRCh38
rs753280770 Chr.4: 87980029-87980029 on Build GRCh38
rs143446619 Chr.4: 87980073-87980073 on Build GRCh38
rs140258871 Chr.4: 87980097-87980097 on Build GRCh38
rs753326509 Chr.4: 87980024-87980024 on Build GRCh38
rs139555315 Chr.4: 87980045-87980045 on Build GRCh38
rs576723090 Chr.4: 87979982-87979982 on Build GRCh38
rs779410973 Chr.4: 87980009-87980009 on Build GRCh38
rs1034479103 Chr.4: 87980088-87980088 on Build GRCh38
rs1005080487 Chr.4: 87980040-87980040 on Build GRCh38
rs750622416 Chr.4: 87980071-87980071 on Build GRCh38
rs188523569 Chr.4: 87980106-87980106 on Build GRCh38
rs150066912 Chr.4: 87980098-87980098 on Build GRCh38
rs759961254 Chr.4: 87980025-87980025 on Build GRCh38
rs376874431 Chr.4: 87980004-87980004 on Build GRCh38
rs61731005 Chr.4: 87980078-87980078 on Build GRCh38
rs761507865 Chr.4: 87980008-87980008 on Build GRCh38
rs765472589 Chr.4: 87980028-87980028 on Build GRCh38
rs767865468 Chr.4: 87980069-87980069 on Build GRCh38
rs1011223303 Chr.4: 87980121-87980121 on Build GRCh38

View Full Product Details