Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 54748596-54748869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD4A
Entrez Gene ID
23034
Gene Name
sterile alpha motif domain containing 4A
Gene Aliases
SAMD4, SMAUG, SMAUG1, SMG, SMGA
UniGene
Hs.733324
Gene Chromosome Location
Chr.14: 54567612-54793315 on Build GRCh38
Transcripts
NM_001161576.2 NM_015589.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
956313 c.1021G>A p.A341T Chr.14: 54748859-54748859 on Build GRCh38
1193769 c.978T>C p.D326D Chr.14: 54748816-54748816 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs886487259 Chr.14: 54748749-54748749 on Build GRCh38
rs531193869 Chr.14: 54748632-54748632 on Build GRCh38
rs115404102 Chr.14: 54748795-54748795 on Build GRCh38
rs1002913409 Chr.14: 54748699-54748700 on Build GRCh38
rs749681832 Chr.14: 54748676-54748676 on Build GRCh38
rs551102607 Chr.14: 54748692-54748692 on Build GRCh38
rs576105261 Chr.14: 54748680-54748680 on Build GRCh38
rs1007915568 Chr.14: 54748681-54748681 on Build GRCh38
rs766039542 Chr.14: 54748842-54748842 on Build GRCh38
rs950432034 Chr.14: 54748777-54748777 on Build GRCh38
rs1017501494 Chr.14: 54748691-54748691 on Build GRCh38
rs536937714 Chr.14: 54748621-54748621 on Build GRCh38
rs905383263 Chr.14: 54748623-54748623 on Build GRCh38
rs1009574955 Chr.14: 54748673-54748673 on Build GRCh38
rs534033735 Chr.14: 54748786-54748786 on Build GRCh38
rs368122696 Chr.14: 54748816-54748816 on Build GRCh38
rs375476232 Chr.14: 54748807-54748807 on Build GRCh38
rs997329332 Chr.14: 54748723-54748723 on Build GRCh38
rs187175430 Chr.14: 54748719-54748719 on Build GRCh38
rs867918298 Chr.14: 54748641-54748641 on Build GRCh38
rs761863481 Chr.14: 54748792-54748792 on Build GRCh38
rs753804150 Chr.14: 54748788-54748788 on Build GRCh38
rs1004969429 Chr.14: 54748762-54748762 on Build GRCh38
rs892521097 Chr.14: 54748672-54748672 on Build GRCh38
rs551349853 Chr.14: 54748707-54748707 on Build GRCh38
rs1026027563 Chr.14: 54748776-54748776 on Build GRCh38
rs966288463 Chr.14: 54748699-54748699 on Build GRCh38
rs556236594 Chr.14: 54748661-54748661 on Build GRCh38

View Full Product Details