Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 175356252-175356525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNR
Entrez Gene ID
7143
Gene Name
tenascin R
Gene Aliases
TN-R
UniGene
Hs.659864
Gene Chromosome Location
Chr.1: 175320781-175743702 on Build GRCh38
Transcripts
NM_003285.2 XM_011509949.2 XM_017002218.1 XM_017002219.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
277756 c.3019C>T p.R1007W Chr.1: 175356418-175356418 on Build GRCh38
1336437 c.3048C>A p.H1016Q Chr.1: 175356389-175356389 on Build GRCh38
165048 c.2988C>A p.T996T Chr.1: 175356449-175356449 on Build GRCh38
386179 c.3097A>T p.I1033F Chr.1: 175356340-175356340 on Build GRCh38
3980062 c.3006C>A p.V1002V Chr.1: 175356431-175356431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs900313664 Chr.1: 175356286-175356286 on Build GRCh38
rs761582604 Chr.1: 175356341-175356341 on Build GRCh38
rs139914165 Chr.1: 175356433-175356433 on Build GRCh38
rs770742683 Chr.1: 175356297-175356297 on Build GRCh38
rs757551606 Chr.1: 175356484-175356484 on Build GRCh38
rs181173905 Chr.1: 175356436-175356436 on Build GRCh38
rs184466076 Chr.1: 175356292-175356292 on Build GRCh38
rs373156400 Chr.1: 175356381-175356381 on Build GRCh38
rs139871740 Chr.1: 175356450-175356450 on Build GRCh38
rs200053413 Chr.1: 175356452-175356452 on Build GRCh38
rs766368531 Chr.1: 175356360-175356360 on Build GRCh38
rs139164267 Chr.1: 175356362-175356362 on Build GRCh38
rs779343064 Chr.1: 175356379-175356379 on Build GRCh38
rs748527179 Chr.1: 175356293-175356293 on Build GRCh38
rs761220590 Chr.1: 175356300-175356300 on Build GRCh38
rs777066098 Chr.1: 175356403-175356403 on Build GRCh38
rs377534855 Chr.1: 175356295-175356295 on Build GRCh38
rs777809906 Chr.1: 175356461-175356461 on Build GRCh38
rs767333852 Chr.1: 175356348-175356348 on Build GRCh38
rs756103916 Chr.1: 175356357-175356357 on Build GRCh38
rs200993334 Chr.1: 175356371-175356371 on Build GRCh38
rs772479959 Chr.1: 175356294-175356294 on Build GRCh38
rs148153913 Chr.1: 175356380-175356380 on Build GRCh38
rs771493093 Chr.1: 175356398-175356398 on Build GRCh38
rs753820758 Chr.1: 175356447-175356447 on Build GRCh38
rs762401732 Chr.1: 175356309-175356309 on Build GRCh38
rs756208713 Chr.1: 175356279-175356279 on Build GRCh38
rs766421633 Chr.1: 175356441-175356441 on Build GRCh38
rs370440485 Chr.1: 175356498-175356498 on Build GRCh38
rs868254152 Chr.1: 175356424-175356424 on Build GRCh38
rs547494051 Chr.1: 175356307-175356307 on Build GRCh38
rs747416073 Chr.1: 175356469-175356469 on Build GRCh38
rs747296225 Chr.1: 175356395-175356395 on Build GRCh38
rs769229714 Chr.1: 175356304-175356304 on Build GRCh38
rs147278578 Chr.1: 175356492-175356492 on Build GRCh38
rs867038686 Chr.1: 175356460-175356460 on Build GRCh38
rs758678378 Chr.1: 175356456-175356456 on Build GRCh38
rs750363887 Chr.1: 175356350-175356350 on Build GRCh38
rs77426583 Chr.1: 175356280-175356280 on Build GRCh38
rs978531773 Chr.1: 175356425-175356425 on Build GRCh38
rs769017834 Chr.1: 175356289-175356289 on Build GRCh38
rs774052903 Chr.1: 175356418-175356418 on Build GRCh38
rs146606650 Chr.1: 175356319-175356319 on Build GRCh38
rs112059746 Chr.1: 175356417-175356417 on Build GRCh38
rs566266805 Chr.1: 175356281-175356281 on Build GRCh38
rs538886908 Chr.1: 175356437-175356437 on Build GRCh38
rs773930890 Chr.1: 175356340-175356340 on Build GRCh38
rs1023823430 Chr.1: 175356431-175356431 on Build GRCh38
rs749338933 Chr.1: 175356283-175356283 on Build GRCh38
rs761508521 Chr.1: 175356427-175356427 on Build GRCh38
rs752902192 Chr.1: 175356453-175356453 on Build GRCh38
rs1018011025 Chr.1: 175356376-175356376 on Build GRCh38
rs748804211 Chr.1: 175356413-175356413 on Build GRCh38
rs916636789 Chr.1: 175356488-175356488 on Build GRCh38
rs781314869 Chr.1: 175356344-175356344 on Build GRCh38
rs188781558 Chr.1: 175356387-175356387 on Build GRCh38

View Full Product Details