Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51975296-51975545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1B
Entrez Gene ID
91
Gene Name
activin A receptor type 1B
Gene Aliases
ACTRIB, ACVRLK4, ALK4, SKR2
UniGene
Hs.438918
Gene Chromosome Location
Chr.12: 51951667-51997079 on Build GRCh38
Transcripts
NM_004302.4 NM_020327.3 NM_020328.3 XM_011538966.2 XM_011538967.2 XM_017020199.1 XM_017020200.1 XM_017020201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1128564 c.170T>G p.F57C Chr.12: 51975343-51975343 on Build GRCh38
6229334 c.300C>G p.Y100* Chr.12: 51975473-51975473 on Build GRCh38
5630684 c.188A>T p.E63V Chr.12: 51975361-51975361 on Build GRCh38
5794565 c.189G>T p.E63D Chr.12: 51975362-51975362 on Build GRCh38
4042763 c.206G>A p.C69Y Chr.12: 51975379-51975379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143486670 Chr.12: 51975326-51975326 on Build GRCh38
rs528922488 Chr.12: 51975401-51975401 on Build GRCh38
rs761131576 Chr.12: 51975352-51975352 on Build GRCh38
rs2242106 Chr.12: 51975516-51975516 on Build GRCh38
rs748742473 Chr.12: 51975316-51975316 on Build GRCh38
rs779113347 Chr.12: 51975497-51975497 on Build GRCh38
rs758937576 Chr.12: 51975392-51975392 on Build GRCh38
rs751308109 Chr.12: 51975444-51975444 on Build GRCh38
rs374726820 Chr.12: 51975524-51975524 on Build GRCh38
rs953028798 Chr.12: 51975458-51975458 on Build GRCh38
rs533764202 Chr.12: 51975522-51975522 on Build GRCh38
rs764850274 Chr.12: 51975404-51975404 on Build GRCh38
rs755003561 Chr.12: 51975488-51975488 on Build GRCh38
rs920805019 Chr.12: 51975341-51975341 on Build GRCh38
rs997364413 Chr.12: 51975376-51975376 on Build GRCh38
rs748177354 Chr.12: 51975503-51975503 on Build GRCh38
rs114638064 Chr.12: 51975407-51975407 on Build GRCh38
rs757077272 Chr.12: 51975445-51975445 on Build GRCh38
rs754112316 Chr.12: 51975485-51975485 on Build GRCh38
rs770655746 Chr.12: 51975383-51975383 on Build GRCh38
rs375781164 Chr.12: 51975449-51975449 on Build GRCh38
rs776453681 Chr.12: 51975389-51975389 on Build GRCh38
rs762882277 Chr.12: 51975408-51975408 on Build GRCh38
rs543085357 Chr.12: 51975434-51975434 on Build GRCh38
rs905508945 Chr.12: 51975388-51975388 on Build GRCh38
rs766863860 Chr.12: 51975426-51975426 on Build GRCh38
rs899811006 Chr.12: 51975366-51975366 on Build GRCh38
rs372410454 Chr.12: 51975521-51975521 on Build GRCh38
rs772569915 Chr.12: 51975511-51975511 on Build GRCh38
rs773641366 Chr.12: 51975330-51975330 on Build GRCh38
rs938359774 Chr.12: 51975399-51975399 on Build GRCh38
rs200240312 Chr.12: 51975500-51975500 on Build GRCh38

View Full Product Details