Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28749441-28749694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAF4
Entrez Gene ID
9618
Gene Name
TNF receptor associated factor 4
Gene Aliases
CART1, MLN62, RNF83
UniGene
Hs.8375
Gene Chromosome Location
Chr.17: 28744003-28750958 on Build GRCh38
Transcripts
NM_004295.3 XM_011525504.2 XM_011525505.2 XM_011525506.2 XM_011525507.2 XM_011525508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417009 c.1337G>A p.R446Q Chr.17: 28749501-28749501 on Build GRCh38
238607 c.1343G>A p.R448Q Chr.17: 28749507-28749507 on Build GRCh38
977018 c.1354C>T p.R452W Chr.17: 28749518-28749518 on Build GRCh38
977017 c.1343G>T p.R448L Chr.17: 28749507-28749507 on Build GRCh38
1302570 c.1387G>A p.E463K Chr.17: 28749551-28749551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370031471 Chr.17: 28749559-28749559 on Build GRCh38
rs371664470 Chr.17: 28749593-28749593 on Build GRCh38
rs749558918 Chr.17: 28749530-28749530 on Build GRCh38
rs774977454 Chr.17: 28749566-28749566 on Build GRCh38
rs748249974 Chr.17: 28749547-28749547 on Build GRCh38
rs781202637 Chr.17: 28749514-28749514 on Build GRCh38
rs761110160 Chr.17: 28749491-28749491 on Build GRCh38
rs755843596 Chr.17: 28749613-28749613 on Build GRCh38
rs778922671 Chr.17: 28749540-28749540 on Build GRCh38
rs756872921 Chr.17: 28749621-28749622 on Build GRCh38
rs768689325 Chr.17: 28749534-28749534 on Build GRCh38
rs751150016 Chr.17: 28749507-28749507 on Build GRCh38
rs561636944 Chr.17: 28749580-28749580 on Build GRCh38
rs931836471 Chr.17: 28749557-28749557 on Build GRCh38
rs956278161 Chr.17: 28749668-28749668 on Build GRCh38
rs763740569 Chr.17: 28749501-28749501 on Build GRCh38
rs1048780596 Chr.17: 28749494-28749494 on Build GRCh38
rs757216557 Chr.17: 28749511-28749511 on Build GRCh38
rs892468007 Chr.17: 28749654-28749654 on Build GRCh38
rs1019147464 Chr.17: 28749548-28749548 on Build GRCh38
rs559615823 Chr.17: 28749639-28749639 on Build GRCh38
rs367932681 Chr.17: 28749592-28749592 on Build GRCh38
rs780332977 Chr.17: 28749623-28749651 on Build GRCh38
rs752459129 Chr.17: 28749496-28749496 on Build GRCh38
rs750118646 Chr.17: 28749519-28749519 on Build GRCh38
rs202028805 Chr.17: 28749508-28749508 on Build GRCh38
rs771382560 Chr.17: 28749563-28749563 on Build GRCh38
rs764726724 Chr.17: 28749492-28749492 on Build GRCh38
rs984899858 Chr.17: 28749481-28749481 on Build GRCh38
rs781437530 Chr.17: 28749605-28749605 on Build GRCh38
rs35760160 Chr.17: 28749606-28749606 on Build GRCh38
rs779937752 Chr.17: 28749621-28749621 on Build GRCh38
rs569302687 Chr.17: 28749560-28749560 on Build GRCh38
rs760940325 Chr.17: 28749561-28749561 on Build GRCh38
rs780127506 Chr.17: 28749520-28749520 on Build GRCh38
rs1013557174 Chr.17: 28749663-28749663 on Build GRCh38
rs1032981083 Chr.17: 28749637-28749637 on Build GRCh38
rs763865696 Chr.17: 28749581-28749581 on Build GRCh38

View Full Product Details