Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119629647-119629918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM233
Entrez Gene ID
387890
Gene Name
transmembrane protein 233
Gene Aliases
DSPB2, IFITMD2
UniGene
Hs.530965
Gene Chromosome Location
Chr.12: 119592123-119654455 on Build GRCh38
Transcripts
NM_001136534.1 XM_005253880.3 XM_005253881.3 XM_011538328.2 XM_011538329.2 XM_011538330.2 XM_011538331.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6371751 c.248A>G p.K83R Chr.12: 119629797-119629797 on Build GRCh38
4440848 c.217G>A p.E73K Chr.12: 119629766-119629766 on Build GRCh38
4984359 c.230G>A p.R77Q Chr.12: 119629779-119629779 on Build GRCh38
4830178 c.209G>C p.G70A Chr.12: 119629758-119629758 on Build GRCh38
6347370 c.302G>T p.C101F Chr.12: 119629851-119629851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777520610 Chr.12: 119629779-119629779 on Build GRCh38
rs764198454 Chr.12: 119629824-119629824 on Build GRCh38
rs187625128 Chr.12: 119629838-119629838 on Build GRCh38
rs755987884 Chr.12: 119629778-119629778 on Build GRCh38
rs757010705 Chr.12: 119629788-119629788 on Build GRCh38
rs371582871 Chr.12: 119629797-119629797 on Build GRCh38
rs748460598 Chr.12: 119629864-119629864 on Build GRCh38
rs745431029 Chr.12: 119629811-119629811 on Build GRCh38
rs893946632 Chr.12: 119629781-119629781 on Build GRCh38
rs537113870 Chr.12: 119629787-119629787 on Build GRCh38
rs773602139 Chr.12: 119629897-119629897 on Build GRCh38
rs752467663 Chr.12: 119629766-119629766 on Build GRCh38
rs745882718 Chr.12: 119629882-119629882 on Build GRCh38
rs2301653 Chr.12: 119629756-119629756 on Build GRCh38
rs116350058 Chr.12: 119629733-119629733 on Build GRCh38
rs762977471 Chr.12: 119629754-119629754 on Build GRCh38
rs770171674 Chr.12: 119629887-119629887 on Build GRCh38
rs983282733 Chr.12: 119629895-119629895 on Build GRCh38
rs950639378 Chr.12: 119629878-119629878 on Build GRCh38
rs904374166 Chr.12: 119629710-119629710 on Build GRCh38
rs765383012 Chr.12: 119629722-119629722 on Build GRCh38
rs534588747 Chr.12: 119629692-119629692 on Build GRCh38
rs762234288 Chr.12: 119629693-119629693 on Build GRCh38
rs973288311 Chr.12: 119629810-119629810 on Build GRCh38
rs1003663517 Chr.12: 119629835-119629835 on Build GRCh38
rs992540232 Chr.12: 119629888-119629888 on Build GRCh38
rs973290896 Chr.12: 119629856-119629856 on Build GRCh38
rs375418332 Chr.12: 119629765-119629765 on Build GRCh38
rs777015322 Chr.12: 119629852-119629852 on Build GRCh38
rs911765992 Chr.12: 119629896-119629896 on Build GRCh38
rs754928904 Chr.12: 119629762-119629762 on Build GRCh38
rs576666159 Chr.12: 119629841-119629841 on Build GRCh38
rs751638814 Chr.12: 119629757-119629757 on Build GRCh38
rs1044254021 Chr.12: 119629677-119629677 on Build GRCh38

View Full Product Details