Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48186844-48187117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC11
Entrez Gene ID
85320
Gene Name
ATP binding cassette subfamily C member 11
Gene Aliases
EWWD, MRP8, WW
UniGene
Hs.652267
Gene Chromosome Location
Chr.16: 48164842-48249973 on Build GRCh38
Transcripts
NM_032583.3 NM_033151.3 NM_145186.2 XM_011523397.1 XM_011523398.2 XM_017023795.1 XM_017023796.1 XM_017023797.1 XM_017023798.1 XM_017023799.1 XM_017023800.1 XM_017023801.1 XM_017023802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363093 c.3061T>C p.F1021L Chr.16: 48186963-48186963 on Build GRCh38
6447841 c.2970G>A p.L990L Chr.16: 48187054-48187054 on Build GRCh38
6590312 c.2990C>T p.P997L Chr.16: 48187034-48187034 on Build GRCh38
1165816 c.2953G>A p.G985S Chr.16: 48187071-48187071 on Build GRCh38
5929630 c.3058G>A p.D1020N Chr.16: 48186966-48186966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766414699 Chr.16: 48187002-48187002 on Build GRCh38
rs774189160 Chr.16: 48186985-48186985 on Build GRCh38
rs761817185 Chr.16: 48187059-48187079 on Build GRCh38
rs753061671 Chr.16: 48186947-48186947 on Build GRCh38
rs748870332 Chr.16: 48187074-48187074 on Build GRCh38
rs973480626 Chr.16: 48187086-48187086 on Build GRCh38
rs769455299 Chr.16: 48186967-48186967 on Build GRCh38
rs968927646 Chr.16: 48186887-48186887 on Build GRCh38
rs764204266 Chr.16: 48187072-48187072 on Build GRCh38
rs753039960 Chr.16: 48187038-48187038 on Build GRCh38
rs766539786 Chr.16: 48186915-48186915 on Build GRCh38
rs754089651 Chr.16: 48186923-48186923 on Build GRCh38
rs754590465 Chr.16: 48187089-48187089 on Build GRCh38
rs935756776 Chr.16: 48186896-48186896 on Build GRCh38
rs765156837 Chr.16: 48187078-48187080 on Build GRCh38
rs778440563 Chr.16: 48187091-48187091 on Build GRCh38
rs778150422 Chr.16: 48186965-48186965 on Build GRCh38
rs779816745 Chr.16: 48186970-48186970 on Build GRCh38
rs761021001 Chr.16: 48187041-48187041 on Build GRCh38
rs771985842 Chr.16: 48186997-48186997 on Build GRCh38
rs368123925 Chr.16: 48186911-48186911 on Build GRCh38
rs760757714 Chr.16: 48186914-48186914 on Build GRCh38
rs145321251 Chr.16: 48186999-48186999 on Build GRCh38
rs748923651 Chr.16: 48186980-48186980 on Build GRCh38
rs758939546 Chr.16: 48186959-48186959 on Build GRCh38
rs752147564 Chr.16: 48187066-48187066 on Build GRCh38
rs747926334 Chr.16: 48187096-48187096 on Build GRCh38
rs757882968 Chr.16: 48187071-48187071 on Build GRCh38
rs894131318 Chr.16: 48186956-48186956 on Build GRCh38
rs760705007 Chr.16: 48187001-48187001 on Build GRCh38
rs755234174 Chr.16: 48186928-48186928 on Build GRCh38
rs377357697 Chr.16: 48186888-48186888 on Build GRCh38
rs753231393 Chr.16: 48186940-48186940 on Build GRCh38
rs759741868 Chr.16: 48187011-48187011 on Build GRCh38
rs772002669 Chr.16: 48186907-48186907 on Build GRCh38
rs775424093 Chr.16: 48186936-48186936 on Build GRCh38
rs59029650 Chr.16: 48187040-48187040 on Build GRCh38
rs200790855 Chr.16: 48187007-48187007 on Build GRCh38
rs546792480 Chr.16: 48186890-48186890 on Build GRCh38
rs761515169 Chr.16: 48186992-48186992 on Build GRCh38
rs765445951 Chr.16: 48187020-48187020 on Build GRCh38
rs370070344 Chr.16: 48186981-48186981 on Build GRCh38
rs1020119131 Chr.16: 48186885-48186885 on Build GRCh38
rs1043890105 Chr.16: 48186881-48186881 on Build GRCh38

View Full Product Details