Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37976771-37977044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SF3A3
Entrez Gene ID
10946
Gene Name
splicing factor 3a subunit 3
Gene Aliases
PRP9, PRPF9, SAP61, SF3a60
UniGene
Hs.77897
Gene Chromosome Location
Chr.1: 37956975-37990110 on Build GRCh38
Transcripts
NM_001320830.1 NM_006802.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5632232 c.967C>T p.L323L Chr.1: 37976922-37976922 on Build GRCh38
7389505 c.997A>T p.I333F Chr.1: 37976892-37976892 on Build GRCh38
1470221 c.976C>A p.Q326K Chr.1: 37976913-37976913 on Build GRCh38
6397525 c.945A>T p.E315D Chr.1: 37976944-37976944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530859034 Chr.1: 37976808-37976808 on Build GRCh38
rs982747872 Chr.1: 37976950-37976950 on Build GRCh38
rs368358662 Chr.1: 37976997-37976997 on Build GRCh38
rs954655474 Chr.1: 37976911-37976911 on Build GRCh38
rs756698412 Chr.1: 37976986-37976986 on Build GRCh38
rs748299079 Chr.1: 37976850-37976850 on Build GRCh38
rs769879507 Chr.1: 37976859-37976859 on Build GRCh38
rs774699773 Chr.1: 37976878-37976878 on Build GRCh38
rs570631596 Chr.1: 37976930-37976930 on Build GRCh38
rs10890216 Chr.1: 37976875-37976875 on Build GRCh38
rs767990214 Chr.1: 37976908-37976908 on Build GRCh38
rs763728166 Chr.1: 37976952-37976952 on Build GRCh38
rs753427956 Chr.1: 37976968-37976968 on Build GRCh38
rs771511437 Chr.1: 37976877-37976877 on Build GRCh38
rs760004437 Chr.1: 37976896-37976896 on Build GRCh38
rs963429181 Chr.1: 37976821-37976821 on Build GRCh38
rs762690381 Chr.1: 37976869-37976872 on Build GRCh38
rs760354372 Chr.1: 37976939-37976939 on Build GRCh38
rs969670969 Chr.1: 37976925-37976925 on Build GRCh38
rs908095056 Chr.1: 37977015-37977015 on Build GRCh38
rs781455699 Chr.1: 37976849-37976849 on Build GRCh38
rs766174354 Chr.1: 37976875-37976875 on Build GRCh38
rs921954156 Chr.1: 37976900-37976900 on Build GRCh38

View Full Product Details