Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97003949-97004188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLIT1
Entrez Gene ID
6585
Gene Name
slit guidance ligand 1
Gene Aliases
MEGF4, SLIL1, SLIT-1, SLIT3
UniGene
Hs.632082
Gene Chromosome Location
Chr.10: 96998038-97185926 on Build GRCh38
Transcripts
NM_003061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1289436 c.3835C>T p.L1279F Chr.10: 97004098-97004098 on Build GRCh38
4545295 c.3750G>A p.E1250E Chr.10: 97004183-97004183 on Build GRCh38
3868212 c.3759C>T p.A1253A Chr.10: 97004174-97004174 on Build GRCh38
4545568 c.3810G>A p.M1270I Chr.10: 97004123-97004123 on Build GRCh38
5518994 c.3793G>A p.G1265R Chr.10: 97004140-97004140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927033118 Chr.10: 97004030-97004030 on Build GRCh38
rs898164218 Chr.10: 97004001-97004001 on Build GRCh38
rs753670627 Chr.10: 97004041-97004041 on Build GRCh38
rs145607290 Chr.10: 97004141-97004141 on Build GRCh38
rs753724001 Chr.10: 97004134-97004134 on Build GRCh38
rs752194553 Chr.10: 97004100-97004100 on Build GRCh38
rs774613268 Chr.10: 97004089-97004089 on Build GRCh38
rs374108509 Chr.10: 97004017-97004017 on Build GRCh38
rs763920890 Chr.10: 97004031-97004031 on Build GRCh38
rs764610984 Chr.10: 97004161-97004161 on Build GRCh38
rs760387535 Chr.10: 97004028-97004028 on Build GRCh38
rs757875768 Chr.10: 97004053-97004053 on Build GRCh38
rs760748912 Chr.10: 97004107-97004107 on Build GRCh38
rs778170165 Chr.10: 97004075-97004075 on Build GRCh38
rs778676445 Chr.10: 97004048-97004048 on Build GRCh38
rs772155256 Chr.10: 97004099-97004099 on Build GRCh38
rs527727075 Chr.10: 97003984-97003984 on Build GRCh38
rs749650797 Chr.10: 97004084-97004084 on Build GRCh38
rs371979178 Chr.10: 97004117-97004117 on Build GRCh38
rs752635293 Chr.10: 97004022-97004022 on Build GRCh38
rs759559857 Chr.10: 97004090-97004090 on Build GRCh38
rs1049479639 Chr.10: 97004016-97004016 on Build GRCh38
rs775298979 Chr.10: 97004104-97004104 on Build GRCh38
rs996587206 Chr.10: 97004012-97004012 on Build GRCh38
rs757035233 Chr.10: 97004044-97004044 on Build GRCh38
rs770157444 Chr.10: 97004069-97004069 on Build GRCh38
rs764818906 Chr.10: 97004140-97004140 on Build GRCh38
rs199794015 Chr.10: 97004135-97004135 on Build GRCh38
rs552104302 Chr.10: 97004049-97004049 on Build GRCh38
rs368125888 Chr.10: 97004060-97004060 on Build GRCh38
rs1008374396 Chr.10: 97004043-97004043 on Build GRCh38
rs771185345 Chr.10: 97004085-97004085 on Build GRCh38
rs1001273121 Chr.10: 97004106-97004106 on Build GRCh38
rs764119481 Chr.10: 97004131-97004131 on Build GRCh38
rs896628252 Chr.10: 97004083-97004083 on Build GRCh38
rs375179079 Chr.10: 97004147-97004147 on Build GRCh38
rs779559258 Chr.10: 97004153-97004153 on Build GRCh38

View Full Product Details